Skip to Content
Merck
All Photos(1)

Documents

1.03830

Supelco

HPLC-bottle adapter with 3 tube connections ID 3.2 mm, solvents supply

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121516
NACRES:
NA.03

storage temp.

2-30°C

Related Categories

General description

Safe, easy and contamination free solvent handling is one of the main important topics in daily lab work. Our bottle adapter with included filter-valve connects your S40-threaded glass bottles directly to instruments with 3mm tubes. This avoids escaping solvent vapours as well as contaminations of the solvent to protect your health, the environment and the solvent quality. The specially selected materials provide highest safety and maximum protection against solvent contamination.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service