Skip to Content
Merck
All Photos(1)

Key Documents

1.67206

Supelco

Bottle adapter (PTFE), S40 (bottle thread) to GL45 (outer thread)

Synonym(s):

PTFE Bottle Connector S40 to GL45

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€53.50

€53.50


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
1 UNIT
€53.50

About This Item

UNSPSC Code:
12352203
NACRES:
NB.51

€53.50


Estimated to ship on21 May 2025


Request a Bulk Order

storage temp.

no temp limit

General description

Special S40 bottle adapters made of PTFE provide all possible connection options for safe and reliable daily solvents handling. This adapter is equipped with S40 inner thread and GL45 outer thread.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service