Skip to Content
Merck
All Photos(3)

Key Documents

1.02930

Supelco

1,2-Dichlorobenzene

for extraction analysis EMSURE®

Synonym(s):

1,2-Dichlorobenzene, o-Chlorobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€94.20
2.5 L
€161.00

€94.20


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
€94.20
2.5 L
€161.00

About This Item

Linear Formula:
1,2-(Cl)2C6H4
CAS Number:
Molecular Weight:
147.00
MDL number:
UNSPSC Code:
12191501
EC Index Number:
202-425-9
NACRES:
NA.21
grade:
reagent grade
bp:
180 °C/1013 hPa
vapor pressure:
1.33 hPa ( 20 °C)

€94.20


Estimated to ship on26 May 2025


Request a Bulk Order

grade

reagent grade

Quality Level

vapor pressure

1.33 hPa ( 20 °C)

product line

EMSURE®

Assay

≥99% (GC)

form

liquid

autoignition temp.

640 °C

potency

500 mg/kg LD50, oral (Rat)
>10000 mg/kg LD50, skin (Rabbit)

expl. lim.

2.2-12 % (v/v)

dilution

(for analytical testing)

impurities

≤0.0001% Chloride (Cl)
≤0.0001% Fe (Iron)
≤0.0001% Heavy metals (as Pb)
≤0.001% Free acid (as HCl)
≤0.005% Sulfated ash
≤0.01% Water

bp

180 °C/1013 hPa

mp

-17 °C

transition temp

flash point 66 °C

solubility

0.13 g/L

density

1.31 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Clc1c(cccc1)Cl

InChI

1S/C6H4Cl2/c7-5-3-1-2-4-6(5)8/h1-4H

InChI key

RFFLAFLAYFXFSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Linkage

Discover EMSURE - to be more than sure!

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service