Skip to Content
Merck
All Photos(1)

Key Documents

8.03238

Sigma-Aldrich

1,2-Dichlorobenzene

for synthesis

Synonym(s):

1,2-Dichlorobenzene, o-Chlorobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€21.70
1 L
€57.80
2.5 L
€82.40

€21.70


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€21.70
1 L
€57.80
2.5 L
€82.40

About This Item

Linear Formula:
1,2-(Cl)2C6H4
CAS Number:
Molecular Weight:
147.00
MDL number:
UNSPSC Code:
12352101
EC Index Number:
202-425-9
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
180 °C/1013 hPa
vapor pressure:
1.33 hPa ( 20 °C)

€21.70


Estimated to ship on28 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

1.33 hPa ( 20 °C)

Assay

≥98% (GC)

form

liquid

autoignition temp.

640 °C

potency

500 mg/kg LD50, oral (Rat)
>10000 mg/kg LD50, skin (Rabbit)

expl. lim.

2.2-12 % (v/v)

dilution

(for synthesis)

bp

180 °C/1013 hPa

mp

-17 °C

transition temp

flash point 66 °C

solubility

0.13 g/L

density

1.31 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Clc1c(cccc1)Cl

InChI

1S/C6H4Cl2/c7-5-3-1-2-4-6(5)8/h1-4H

InChI key

RFFLAFLAYFXFSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2-Dichlorobenzene for synthesis. CAS 95-50-1, chemical formula 1,2-(Cl)₂C₆H₄.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.305 - 1.307
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service