Skip to Content
Merck

Skip To

1.02440

Parteck® Mg DC (Magnesium hydroxide carbonate) heavy

EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, USP, E 504

Synonym(s):

Magnesium carbonate basic

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kg
Check Cart for Availability
€ 249,00
40 kg
Check Cart for Availability
€ 7.640,00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352107
E Number:
E504
EC Index Number:
235-192-7

€ 249,00


Check Cart for Availability

Request a Bulk OrderRequest a Sample
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, JP, Ph. Eur., USP

product line

EMPROVE® ESSENTIAL

form

solid

composition

Mg (as MgO), 40.0-43.5% alkalimetric (USP), Mg (as MgO), 40.0-43.5% complexometric , Mg (as MgO), 40.0-44.0% complexometric (JP)

particle size

21-51 μm (d50)

pH

10.5 (20 °C, 50 g/L in H2O, suspension)

mp

600 °C (decomposition)

solubility

water: insoluble at 20 °C

density

2.16 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤0.02%, nitrate (NO3-): ≤0.015%, nitrite (NO2-): ≤0.015%, sulfate (SO42-): ≤0.3%

cation traces

As: ≤0.0002%, Ca: ≤0.4%, Fe: ≤0.005%, Hg: ≤0.0001%, Na: ≤0.4%, Pb: ≤0.0002%, heavy metals (as Pb): ≤0.001%

application(s)

pharmaceutical
solid formulation

storage temp.

2-30°C

SMILES string

[Mg+2].[O-]C(=O)[O-].[O-H]

InChI

1S/CH2O3.Mg.H2O/c2-1(3)4;;/h(H2,2,3,4);;1H2/q;+2;/p-3

InChI key

OUHCLAKJJGMPSW-UHFFFAOYSA-K

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Parteck® Mg DC is mainly used in highly regulated pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
PARTECK is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.058671.371821.02064
application(s)

pharmaceutical
solid formulation

application(s)

pharmaceutical
solid formulation

application(s)

liquid formulation
pharma/biopharma processes
pharmaceutical
solid formulation

application(s)

liquid formulation
pharmaceutical
solid formulation

agency

BP, Ph. Eur., JP, USP

agency

BP, ChP, Ph. Eur., USP

agency

BP, JP, Ph. Eur., USP

agency

BP, JP, Ph. Eur., USP

form

solid

form

solid

form

solid

form

solid

solubility

water: insoluble at 20 °C

solubility

-

solubility

710 g/L

solubility

-

product line

EMPROVE® ESSENTIAL

product line

EMPROVE® ESSENTIAL

product line

EMPROVE® EXPERT

product line

EMPROVE® ESSENTIAL

density

2.16 g/cm3 at 20 °C

density

3.58 g/cm3 at 25 °C

density

1.68 g/cm3 at 20 °C

density

2.93 g/mL at 25 °C (lit.)


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters