Skip to Content
Merck

Skip To

1.02431

Chloroform

EMPROVE® ESSENTIAL

Synonym(s):

TCM, Trichloromethane, Methane trichloride, Methyl trichloride, Methylidyne trichloride, Trichloromethane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 L
Check Cart for Availability
€ 70,60
2.5 L
Check Cart for Availability
€ 130,00
25 L
Check Cart for Availability
€ 561,00
190 L
Check Cart for Availability
€ 2.800,00

About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
EC Index Number:
200-663-8
UNSPSC Code:
12191502
Beilstein/REAXYS Number:
1731042
MDL number:

€ 70,60


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

4.1 (vs air)

Quality Segment

vapor pressure

160 mmHg ( 20 °C)

product line

EMPROVE® ESSENTIAL

assay

99.0-99.4% (GC)

form

liquid

technique(s)

API processing | polymorph screening: suitable

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

density

1.492 g/mL at 25 °C (lit.)

application(s)

liquid formulation
pharmaceutical

storage temp.

2-30°C

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Chloroform is used in pharmaceutical production processes and as an inactivation ingredient in vaccines.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.07024CX1060CX1055
application(s)

liquid formulation
pharmaceutical

application(s)

-

application(s)

-

application(s)

-

technique(s)

API processing | polymorph screening: suitable

technique(s)

-

technique(s)

-

technique(s)

-

form

liquid

form

liquid

form

liquid

form

liquid

assay

99.0-99.4% (GC)

assay

≥99.8% (according to ACS), 99.0-99.4% (GC)

assay

≥99.8% chloroform + ethanol basis

assay

≥99.8% chloroform + ethanol basis

density

1.492 g/mL at 25 °C (lit.)

density

1.492 g/mL at 25 °C (lit.)

density

1.492 g/mL at 25 °C (lit.)

density

1.492 g/mL at 25 °C (lit.)

product line

EMPROVE® ESSENTIAL

product line

EMPARTA®

product line

-

product line

-


Still not finding the right product?

Explore all of our products under Chloroform


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-66-3

CAS No.


Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Instead of addressing API solubility during formulation, tackle it early during development. Read about the various methods for API processing.





Global Trade Item Number

SKUGTIN
102431919004022536033426
102431602404065266024630
102431100004022536033358
102431250004022536033372
102431999804022536716077
102431602504022536033402
102431619004022536905709

Questions

Reviews

No rating value

Active Filters