Product 1024190250 is intended for In Vitro Diagnostic Use, and the Instructions For Use mention staining cryosections but not cultured cells attached to either a culture plate or a chamber slide. Staining cultured cells would be considered "off label" use and would require validation, especially in a clinical lab setting. If the dry dye is used to prepare Oil Red O staining solution, the process for preparing the staining solution would be the same for both cryosections and cells attached to a cell culture plate or chamber slide.
Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
250 mL | Check Cart for Availability | € 135,00 |
About This Item
Quality Segment
form
liquid
IVD
for in vitro diagnostic use
technique(s)
microbe id | staining: suitable
transition temp
flash point -11 °C
density
0.838 g/cm3
application(s)
clinical testing
diagnostic assay manufacturing
hematology
histology
storage temp.
15-25°C
General description
Approx. 200 stains can be performed with one 250 ml bottle. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.
Analysis Note
Lipids: bright red
Nuclei: blue
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) microbe id | staining: suitable | technique(s) microbe id | staining: suitable | technique(s) microbe id | staining: suitable | technique(s) microbe id | staining: suitable |
| form liquid | form liquid | form liquid | form powder |
| application(s) clinical testing | application(s) diagnostic assay manufacturing | application(s) diagnostic assay manufacturing | application(s) diagnostic assay manufacturing |
| storage temp. 15-25°C | storage temp. room temp | storage temp. room temp | storage temp. room temp |
| IVD for in vitro diagnostic use | IVD - | IVD - | IVD - |
| Quality Level 400 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system
supp_hazards
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
12.2 °F
flash_point_c
-11 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Does Oil Red O staining solution 1024190250 also work for cells in culture?
1 answer-
Helpful?
-
-
Need to know whether or not the Oil red O staining solution (1024190250) works in mice liver?
1 answer-
Oil Red O is soluble in water and the solubility test is conducted to 1 mg per ml. The actual maximum solubility can be higher, up to 10 mg/ml, although this may. However, the solubility test is specific to water and may differ if used in agar.
Helpful?
-
-
Is the Propanol in the Oil red O staining solution Halal en Kosher ?
1 answer-
The Oil red O staining solution is prepared with ethanol and acetone. Neither of these components are certified Kosher or Halal. The staining protocol requires 2-propanol. In the product information sheet 1.09634, 2-isopropanol, is suggested. This product is also not certified Halal or Kosher. See the link below to review this protocol:
https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/374/144/102419-en.pdfThe food grade 2-propanol products do offer this certification. See the link below for an example: https://www.sigmaaldrich.com/US/en/product/aldrich/w292907
Helpful?
-
-
Does this work to in mouse liver frozen slices?
1 answer-
Oil Red O lacks the specificity of an antibody. The basic theory of fat or lipid staining is that the dye is dissolved in a solvent with minimal solubility. A working solution is made just prior to use to further decrease the solubility. The dye then moves from the solution with minimal solubility to the solvent where there is better solubility. In this case the dye moves from the solution to the fat or lipid being stained. It matters not whether the tissue is from a human, mouse or other animals.
This information is true to most fat or lipid stains. But with Oil Red O the solution is prepared to a working solution and no further dilution is required.
Helpful?
-



