Skip to Content
Merck

Skip To

1.01910

N,N-Bis(2-hydroxyethyl) glycine

(BICINE) buffer substance

Synonym(s):

N,N-Bis(2-hydroxyethyl) glycine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
250 g
Check Cart for Availability
€ 326,00

About This Item

Empirical Formula (Hill Notation):
C6H13NO4
CAS Number:
Molecular Weight:
163.17
UNSPSC Code:
12161703
EC Index Number:
205-755-1
NACRES:
NA.05
MDL number:

€ 326,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

pH

4-5 (20 °C, 100 g/L in H2O)

mp

185-192 °C

solubility

100 g/L

bulk density

364 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].N(CCO)(CCO)CC(=O)[O-]

InChI

1S/C6H13NO4.Na/c8-3-1-7(2-4-9)5-6(10)11;/h8-9H,1-5H2,(H,10,11);/q;+1/p-1

InChI key

MFBDBXAVPLFMNJ-UHFFFAOYSA-M

Analysis Note

Assay (perchloric acid titration): ≥ 99 %
Identity (IR-spectrum): passes test
UV-Absorption (250 nm, 50 g/l, 1 cm, water): ≤ 0.05
Chloride (Cl): ≤ 0.005 %
Sulphate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
Loss on Drying (105°C): ≤ 0.2 %
Biological buffer, pKa at 20 °C: 8.35

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
391336-MB86601.15228
pH

4-5 (20 °C, 100 g/L in H2O)

pH

-

pH

4.0-6.0 (1 M in H2O)

pH

-

solubility

100 g/L

solubility

water: 5 mg/mL

solubility

H2O: 1 M, clear, colorless

solubility

-

storage temp.

2-30°C

storage temp.

10-30°C

storage temp.

room temp

storage temp.

2-30°C

bulk density

364 kg/m3

bulk density

-

bulk density

-

bulk density

-

mp

185-192 °C

mp

-

mp

-

mp

-

Quality Level

100

Quality Level

100

Quality Level

400

Quality Level

100


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
101910002504022536026091
101910025004022536026107

Questions

Reviews

No rating value

Active Filters