Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
25 tests | Check Cart for Availability | € 198,00 |
About This Item
Product Name
Surfactants (nonion.) Cell Test, photometric, 0.10-7.50 mg/L (Triton™ X-100), Spectroquant®
product line
Spectroquant®
Quality Segment
usage
sufficient for 25 tests
analyte functional class(es)
surfactants, nonionic
measuring range
0.10-7.50 mg/L (Triton™ X-100)
technique(s)
photometry: suitable
application(s)
drinking water
environmental
ground and surface water
wastewater
compatibility
for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)
detection method
photometric (TBPE)
storage temp.
15-25°C
General description
This Spectroquant® Surfactants (nonion.) Cell Test allows the determination nonionic surfactants (hydrophilic end doesn′t carry any charge) in water samples. Surfactants are additives that lower the surface tension of water such as foaming agents, emulsifiers, and detergents. Most surfactants enter the waterways as part of household waste or have their origin in industrial processes and often facilitate the introduction of other contaminates. Enhanced levels of surfactants might cause a hazard for plants, animals, and humans.
The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
Application
</UL>
To monitor water quality in various environments, including rivers, lakes, and wastewater treatment plants. These tests help detect and quantify surfactant levels, ensuring that water
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| detection method photometric (TBPE) | detection method photometric (Methylene blue) | detection method photometric (Disulfine blue) | detection method photometric (Barium sulfate, turbidimetric) |
| technique(s) photometry: suitable | technique(s) photometry: suitable | technique(s) photometry: suitable | technique(s) photometry: suitable |
| measuring range 0.10-7.50 mg/L (Triton™ X-100) | measuring range 0.05 -2.12 mg/L (SDS), 0.05-2.00 mg/L (MBAS), 0.06-2.56 mg/L (SDBS), 0.08-3.26 mg/L (SDOSSA) | measuring range 0.05-1.50 mg/L (CTAB) | measuring range 50-500 mg/L (SO42-) |
| compatibility for use with Spectroquant® Move 100, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 600 (plus), for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 300 (plus) | compatibility for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus) | compatibility for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus) | compatibility for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus) |
| analyte functional class(es) surfactants, nonionic | analyte functional class(es) surfactants, anionic | analyte functional class(es) surfactants (cation) | analyte functional class(es) - |
| storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 15-25°C |
signalword
Warning
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 2 - STOT SE 3
Storage Class
10 - Combustible liquids
target_organs
Central nervous system, Eyes,Central nervous system
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.





