Skip to Content
Merck
All Photos(2)

Key Documents

1.01557

Supelco

Potassium chloride solution

nominal 0.147 mS/cm, certified reference material for the measurement of electrolytic conductivity (c=0.001 mol/l) Certipur®

Synonym(s):

Potassium chloride solution

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€307.00

€307.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
500 ML
€307.00

About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
Beilstein:
1711999
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€307.00


Estimated to ship on29 May 2025


Request a Bulk Order

Product Name

Potassium chloride solution, nominal 0.147 mS/cm, certified reference material for the measurement of electrolytic conductivity (c=0.001 mol/l) Certipur®

grade

certified reference material

Quality Level

Agency

ISO (17025, 17034)

product line

Certipur®

density

0.998 g/cm3 at 20 °C

application(s)

environmental

format

single component solution

storage temp.

15-25°C

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is directly traceable to the corresponding primary reference material from the Physikalisch Technische Bundesanstalt (PTB), Germany. The electrolytic conductivity of this standard is validated using a conductivity meter from an accredited Merck calibration laboratory.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Certified reference material (CRM) for use in aqueous solution for measuring conductivity cell constant or as a control sample.

Analysis Note

Electrical conductivity (25 °C): 0.144 - 0.150 mS/cm

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service