Skip to Content
Merck

Skip To

1.00729

Sulfuric acid

90-91%, for Gerber fat determination and determination of nitrates in milk

Synonym(s):

Sulfuric acid solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
2.5 L
Check Cart for Availability
€ 127,00
25 L
Check Cart for Availability
€ 474,00
€ 355,50

About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
NACRES:
NA.21
UNSPSC Code:
12352301
MDL number:
Beilstein/REAXYS Number:
2037554
Assay:
90.0-91.0% (acidimetric)
Form:
liquid

€ 127,00


Check Cart for Availability
Packaging: Glass bottle

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Sulfuric acid 90-91%, for Gerber fat determination and determination of nitrates in milk

Quality Segment

assay

90.0-91.0% (acidimetric)

form

liquid

impurities

≤0.2 ppm Nitrate (NO3)

bp

300 °C

density

1.81 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): 90.0 - 91.0 %
Density (20 °C): 1.815 - 1.821 g/ml
Nitrate (NO₃): ≤ 0.2 ppm
Test for oily and fatty substances: passes test
Expiry date: see product label

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.007481.007321.09286
assay

90.0-91.0% (acidimetric)

assay

≥97.5% (alkalimetric)

assay

95-97% (alkalimetric)

assay

-

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

no temp limit

storage temp.

no temp limit

storage temp.

no temp limit

storage temp.

15-25°C

density

1.81 g/cm3 at 20 °C

density

1.840 g/mL at 25 °C (lit.)

density

1.840 g/mL at 25 °C (lit.)

density

1.30 g/cm3 at 20 °C

impurities

≤0.2 ppm Nitrate (NO3)

impurities

-

impurities

-

impurities

≤10 ppm Substances reducing potassium permanganate (as SO2)


Still not finding the right product?

Explore all of our products under Sulfuric acid 90-91%


pictograms

Corrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class

8B - Non-combustible, corrosive hazardous materials

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
100729902504022536009834
100729250004022536009810
100729050004022536009803

Questions

Reviews

No rating value

Active Filters