Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 kg | Check Cart for Availability | € 317,00 € 237,75 |
20 kg | Check Cart for Availability | € 5.830,00 |
About This Item
€ 237,75
List Price€ 317,00Save 25%Quality Segment
assay
≥99% SiO2 basis (calculated on ignited on substance, gravimetric)
form
solid
potency
>10000 mg/kg LD50, oral (Rat), >5000 mg/kg LD50, skin (Rabbit)
impurities
≤1% With hydrofluoric acid non volatile matter (as sulfate, ignited basis)
loss
10-20% loss on ignition, 1000 °C
mp
1710 °C
solubility
water: insoluble at 20 °C
density
2-6 g/cm3 at 20 °C
bulk density
60‑600 kg/m3
anion traces
chloride (Cl-): ≤0.05%
cation traces
Fe: ≤0.03%, heavy metals (as Pb): ≤0.005%
application(s)
sample preparation
storage temp.
2-30°C
SMILES string
[Si](=O)=O
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Analysis Note
Assay (gravimetric, SiO₂, calc. on ignited substance): ≥ 99 %
Chloride (Cl): ≤ 0.05 %
Heavy metals (as Pb): ≤ 0.005 %
Fe (Iron): ≤ 0.03 %
With hydrofluoric acid non volatile matter (as sulfate, ignited basis): ≤ 1 %
Loss on ignition (1000 °C): 10 - 20 %
Particle size (< 0.1 mm): about 99 %
Bulk density: 20 - 60 g/100 ml
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥99% SiO2 basis (calculated on ignited on substance, gravimetric) | assay 99-100.5% SiO2 basis (calculated on ignited substance, gravimetric) | assay 99% trace metals basis | assay - |
| Quality Level 200 | Quality Level 200 | Quality Level 100 | Quality Level - |
| form solid | form solid | form solid | form powder |
| solubility water: insoluble at 20 °C | solubility water: insoluble at 20 °C | solubility - | solubility - |
| storage temp. 2-30°C | storage temp. no temp limit | storage temp. - | storage temp. - |
| impurities ≤1% With hydrofluoric acid non volatile matter (as sulfate, ignited basis) | impurities - | impurities - | impurities - |
Still not finding the right product?
Explore all of our products under Silicic acid
Storage Class
13 - Non Combustible Solids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

