Skip to Content
Merck
All Photos(1)

Key Documents

1.00809

Supelco

Fluoride Test, photometric

photometric, 0.10-1.80 mg/L (F-), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

25 TESTS
€133.00

€133.00


Estimated to ship on17 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
25 TESTS
€133.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€133.00


Estimated to ship on17 May 2025


Request a Bulk OrderRequest more information

Product Name

Fluoride Cell Test, photometric, 0.10-1.80 mg/L (F-), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

fluoride

measuring range

0.025-0.500 mg/L (F-)
0.10-1.80 mg/L (F-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Alizarin complexone)

storage temp.

15-25°C

General description

Method: photometric 0.10 - 1.80 mg/l F⁻ Spectroquant®
This Spectroquant® Fluoride test allows the accurate quantification of fluoride ions in various waters. Fluoride ions occur on earth in several minerals, but are only present in trace quantities in bodies of water in nature. In a buffered, weakly acidic solution, fluoride ions react with alizarin complexone and lanthanum(III) to form a violet complex that is determined photometrically.
The method is analogous to EPA 340.3 and APHA 4500-F- E.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
The Spectroquant® Cell Tests come with prefilled 16 mm round cells as well as all the required reagents to perform the analysis according to the procedure described in the accompanying instruction leaflet.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service