Skip to Content
Merck

Skip To

1.00517

Perchloric acid 70%

Suprapur®

Synonym(s):

Perchloric acid, PCA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
250 mL
Check Cart for Availability
€ 376,00
€ 319,60
1 L
Check Cart for Availability
€ 938,00
€ 797,30

About This Item

Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
UNSPSC Code:
12352301
NACRES:
NA.21
MDL number:
Assay:
69-72%
Form:
liquid

€ 319,60

List Price€ 376,00Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

for inorganic trace analysis

assay

69-72%

form

liquid

bp

198.7 °C/1013 hPa

mp

-18 °C

density

1.68 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

OCl(=O)(=O)=O

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)

InChI key

VLTRZXGMWDSKGL-UHFFFAOYSA-N

Analysis Note

  • Assay (acidimetric): 69 – 72%
  • Colour: ≤ 10 Hazen
  • Chloride (Cl): ≤ 1000 ppb
  • Phosphate (PO₄): ≤ 100 ppb
  • Sulphate (SO₄): ≤ 1000 ppb
  • Ag (Silver): ≤ 1 ppb
  • Al (Aluminium): ≤ 5 ppb
  • As (Arsenic): ≤ 0.5 ppb
  • Au (Gold): ≤ 0.5 ppb
  • Ba (Barium): ≤ 1 ppb
  • Be (Beryllium): ≤ 0.5 ppb
  • Bi (Bismuth): ≤ 0.5 ppb
  • Ca (Calcium): ≤ 30 ppb
  • Cd (Cadmium): ≤ 1 ppb
  • Co (Cobalt): ≤ 0.5 ppb
  • Cr (Chromium): ≤ 5.0 ppb
  • Cu (Copper): ≤ 0.5 ppb
  • Fe (Iron): ≤ 20 ppb
  • Ga (Gallium): ≤ 0.5 ppb
  • Ge (Germanium): ≤ 0.5 ppb
  • Hg (Mercury): ≤ 5 ppb
  • In (Indium): ≤ 0.5 ppb
  • K (Potassium): ≤ 5 ppb
  • Li (Lithium): ≤ 0.5 ppb
  • Mg (Magnesium): ≤ 5 ppb
  • Mn (Manganese): ≤ 1 ppb
  • Mo (Molybdenum): ≤ 0.5 ppb
  • Na (Sodium): ≤ 20 ppb
  • Ni (Nickel): ≤ 2 ppb
  • Pb (Lead): ≤ 1 ppb
  • Pd (Palladium): ≤ 1 ppb
  • Pt (Platinum): ≤ 5 ppb
  • Sb (Antimony): ≤ 0.5 ppb
  • Sn (Tin): ≤ 1 ppb
  • Sr (Strontium): ≤ 0.5 ppb
  • Ti (Titanium): ≤ 1 ppb
  • Tl (Thallium): ≤ 0.5 ppb
  • U (Uranium): ≤ 0.5 ppb
  • V (Vanadium): ≤ 0.5 ppb
  • Zn (Zinc): ≤ 5 ppb
  • Zr (Zirconium): ≤ 0.5 ppb
  • Residue on ignition (as sulphate): ≤ 2000 ppb
  • Actual analysis values are subject to unavoidable systematic variations in this concentration range

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.005141.0051830755-M
assay

69-72%

assay

-

assay

-

assay

70.0-72.0%

Quality Level

100

Quality Level

300

Quality Level

300

Quality Level

100

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

-

density

1.68 g/cm3 at 20 °C

density

1.68 g/cm3 at 20 °C

density

1.53 g/cm3 at 15 °C

density

1.670 g/cm3

bp

198.7 °C/1013 hPa

bp

198.7 °C/1013 hPa

bp

-

bp

-


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 1 - Skin Corr. 1A - STOT RE 2

target_organs

Thyroid

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.





Global Trade Item Number

SKUGTIN
100517100004022536007205
100517025004022536007199

Questions

Reviews

No rating value

Active Filters