Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 L | Check Cart for Availability | € 42,60 |
25 L | ||
50 L | Check Cart for Availability | € 9.370,00 |
About This Item
Product Name
Sodium hydroxide solution 10%, EMPROVE® EXPERT
Quality Segment
product line
EMPROVE® EXPERT
assay
9.9-10.1% (acidimetric, NaOH)
form
liquid
pH
13.8 (20 °C in H2O)
density
1.11 g/cm3 at 20 °C
application(s)
cleaning in place
pharma/biopharma processes
storage temp.
no temp limit
General description
As a result, you can integrate our products directly into your biopharmaceutical production process. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs.
Application
Recommended for high-risk applications: All our MQ 500 GMP (IPEC-PQG/ EXCiPACT) solutions for CIP and buffer preparation are manufactured under excipient GMP conditions and feature low endotoxin and bioburden levels.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| form liquid | form liquid | form liquid | form liquid |
| product line EMPROVE® EXPERT | product line EMPROVE® EXPERT | product line EMPROVE® EXPERT | product line EMPROVE® EXPERT |
| Quality Level 500 | Quality Level 500 | Quality Level 500 | Quality Level 500 |
| density 1.11 g/cm3 at 20 °C | density 1.52 g/cm3 at 20 °C | density 1.01 g/cm3 at 20 °C | density 1.28 g/cm3 at 20 °C |
| storage temp. no temp limit | storage temp. 15-25°C | storage temp. no temp limit | storage temp. no temp limit |
| pH 13.8 (20 °C in H2O) | pH 14 (20 °C, 89 g/L in H2O) | pH 13.5 (20 °C in H2O) | pH 14 (20 °C in H2O) |
Still not finding the right product?
Explore all of our products under Sodium hydroxide solution 10%
signalword
Danger
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
wgk
WGK 1
hcodes
Storage Class
8B - Non-combustible corrosive hazardous materials
Stay compliant with GMP using the Emprove® Program
Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.
Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.
Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite
Not finding what you are looking for? Visit the Emprove® Program Help page.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

