Skip to Content
Merck
All Photos(1)

Key Documents

1.00221

SAFC

Sodium hydroxide solution

10 M, SUITABLE FOR CLEANING IN PLACE

Sign Into View Organizational & Contract Pricing

Select a Size

25 L
€244.00
1002219950
Inquire

€244.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
25 L
€244.00
1002219950
Inquire

About This Item

UNSPSC Code:
12352305

€244.00


Estimated to ship on02 June 2025


Request a Bulk Order

Product Name

SODIUM HYDROXIDE SOLUTION 10 MOL/L, SUITABLE FOR CLEANING IN PLACE

Quality Level

form

liquid

pH

14 (20 °C in H2O)

density

1.33 g/cm3 at 20 °C

application(s)

pharma/biopharma processes

storage temp.

2-8°C

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Sodium hydroxide solution 10 mol/L CIP is suitable for cleaning in place use in pharmaceutical processes.

Analysis Note

Assay (acidimetric,NaOH)
9.8 - 10.2 mol/l29.5 - 30.7 %Carbonate (as Na₂CO₃)≤ 0.5 %Chloride (Cl)≤ 0.006 %Phosphate (PO₄)≤ 0.0005 %Silicate (SiO₂)≤ 0.005 %Sulfate (SO₄)≤ 0.003 %Heavy metals (as Pb)≤ 0.0005 %Al (Aluminium)≤ 0.001 %Fe (Iron)≤ 0.001 %

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service