Skip to Content
Merck

1.37023

SAFC

Sodium hydroxide solution

32%, EMPROVE® EXPERT

Pharma Manufacturing

Synonym(s):

Sodium hydroxide solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€58.00
2.5 L
€100.00
5 L
€165.00
25 L
€495.00
200 L
Inquire
950 L
€12,950.00

€58.00


Estimated to ship on28 May 2025


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 L
€58.00
2.5 L
€100.00
5 L
€165.00
25 L
€495.00
200 L
Inquire
950 L
€12,950.00

About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305

€58.00


Estimated to ship on28 May 2025


Request a Bulk OrderRequest a Sample

Product Name

Sodium hydroxide solution 32%, EMPROVE® EXPERT

vapor pressure

3 mmHg ( 37 °C)
8 hPa ( 20 °C)

Quality Level

product line

EMPROVE® EXPERT

form

liquid

pH

14 (20 °C in H2O)

mp

9 °C

density

1.35 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (acidimetric,NaOH): 32.0 - 33.0 %
Carbonate (as Na₂CO₃): ≤ 1.0 %
Chloride (Cl): ≤ 70 ppm
Sulfate (SO₄): ≤ 30 ppm
Heavy metals (as Pb): ≤ 5.0 ppm
Al (Aluminium): ≤ 10 ppm
Fe (Iron): ≤ 4ppm
Ni (Nickel) *: ≤ 2.00 ppm
Residual solvents (ICH Q3C): ...excluded by production process
Filtered with 0.2 µm
This product has been manufactured from Sodium hydroxide pellets Ph Eur, BP, JP, NF and waterfor injection (wfi) acc. USP which conforms with highly purified water (hpw) acc. Ph Eur.
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service