Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 100 μg | Check Cart for Availability | € 339,00 |
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
saturated ammonium sulfate (SAS) precipitated
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
technique(s)
ELISA: suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... PADI4(23569)
General description
Immunogen
Application
Epigenetics & Nuclear Function
Histones
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
HEK293 cell lysate
Western Blotting Analysis: 1:25-1:500 dilution of this antibody detected PADI4 in HEK293 cell lysate.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| antibody form saturated ammonium sulfate (SAS) precipitated | antibody form purified antibody | antibody form affinity isolated antibody | antibody form IgG fraction of antiserum |
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| clone polyclonal | clone 10F1, monoclonal | clone polyclonal | clone polyclonal |
| biological source rabbit | biological source mouse | biological source rabbit | biological source rabbit |
| technique(s) ELISA: suitable, western blot: suitable | technique(s) ChIP: suitable, immunoprecipitation (IP): suitable, western blot: suitable | technique(s) immunoblotting: 0.04-0.4 μg/mL, immunohistochemistry: 1:50-1:200 | technique(s) immunoprecipitation (IP): 2-4 μL using DMSO-treated HL60 cells, western blot: 1:500-1:1,000 using DMSO-treated HL60 cells |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



