Skip to Content
Merck

Skip To

07-862

Anti-phospho-PDGFRα (Tyr754) Antibody

Upstate®, from rabbit

Synonym(s):

CD140A, MGC74795, PDGFR2, platelet-derived growth factor receptor, alpha

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 372,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
mouse, human
Application:
WB
Citations:
1

€ 372,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr754)

Gene Information

human ... PDGFRA(5156)

General description

170-185 kDA
The previously assigned protein identifier Q9UD28 has been merged into P16234. Full details can be found on the UniProt database.

Immunogen

Peptide corresponding to amino acid region encompassing the human phospho-PDGFRα (Tyr754).

Application

Research Category
Signaling
Research Sub Category
Growth Factors & Receptors
This Anti-phospho-PDGFRα (Tyr754) Antibody is validated for use in WB for the detection of phospho-PDGFRα (Tyr754).

Biochem/physiol Actions

Human and Mouse PDGFRα that is phosphorylated on Tyrosine 754 and 762, respectively. Rat is predicted to react based on sequence homology.
Predicted to cross-react with rat based on 100% sequence similarity, but has not been tested.

Physical form

100 μL of affinity purified rabbit polyclonal IgG in buffer (PBS (without Mg2+ and Ca2+), pH 7.4 containing 1.0 mg/mL BSA (IgG, protease free) and 0.05% sodium azide).
Affinity purified

Preparation Note

2 years at -80°C.
Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -80°C. Avoid repeated freeze/ thaw cycles, which may damage IgG and affect product performance.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
07-86107-27607-1436
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

species reactivity

mouse, human

species reactivity

mouse, human

species reactivity

human, mouse

species reactivity

human, mouse

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

Gene Information

human ... PDGFRA(5156)

Gene Information

human ... PDGFRA(5156)

Gene Information

human ... PDGFRA(5156)

Gene Information

human ... PDGFA(5154)

technique(s)

western blot: suitable

technique(s)

western blot: suitable

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

ELISA: suitable, immunohistochemistry: suitable (paraffin), neutralization: suitable, western blot: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Regulation of platelet-derived growth factor receptor function by integrin-associated cell surface transglutaminase.
Zemskov, EA; Loukinova, E; Mikhailenko, I; Coleman, RA; Strickland, DK; Belkin, AM
The Journal of Biological Chemistry null



Global Trade Item Number

SKUGTIN
07-86204053252474422

Questions

Reviews

No rating value

Active Filters