Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
200 μg | Check Cart for Availability | € 319,00 |
About This Item
€ 319,00
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human, mouse
manufacturer/tradename
Upstate®
technique(s)
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... IKBKE(9641)
mouse ... Ikbke(56489)
General description
Immunogen
Application
Signaling
Immunological Signaling
Biochem/physiol Actions
Physical form
Preparation Note
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| clone polyclonal | clone monoclonal | clone polyclonal | clone 1D7, recombinant monoclonal |
| biological source rabbit | biological source mouse | biological source rabbit | biological source rabbit |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| species reactivity human, mouse | species reactivity mouse, human | species reactivity human | species reactivity human, mouse |
| antibody form purified antibody | antibody form purified immunoglobulin | antibody form affinity isolated antibody | antibody form purified antibody |
| technique(s) western blot: suitable | technique(s) western blot: suitable | technique(s) immunohistochemistry: suitable, indirect ELISA: suitable, western blot: suitable | technique(s) immunofluorescence: suitable, immunohistochemistry: suitable, western blot: suitable |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



