Skip to Content
Merck

Skip To

07-307

Anti-phospho-STAT1 (Tyr701) Antibody

Upstate®, from rabbit

Synonym(s):

Transcription factor ISGF-3 components p91/p84, signal transducer and activator of transcription 1, signal transducer and activator of transcription 1, 91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of tr

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 438,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
mouse, chicken, human, rat
Application:
WB
Citations:
25

€ 438,00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1520

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, chicken, human, rat

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr701)

Gene Information

human ... STAT1(6772)

General description

92 kDa
STAT proteins (Signal Transduction and Activators of Transcription) are latent cytoplasmic transcription factors that have the dual function of signal transduction and activation of transcription. STATs are activated by tyrosine phosphorylation in response to different ligands, after which they translocate to the cell nucleus. The N-terminal region is highly homologous among the STAT proteins and surrounds a completely conserved arginine residue. STATs are a part of the JAK-STAT signaling pathway – a major pathway of the immune system. All cytokines transduce critical signals through this pathway. STAT 1, is activated by a number of different ligands, including IFNalpha, IFNgamma, EGF, PDGF and IL6. Phosphorylation of tyrosine 701 is required for STAT 1 dissociation from IFNGR1, homodimerization, and nuclear translocation. Tyrosine 701 phosphorylation impairment results in loss of STAT 1 functions.

Immunogen

Epitope: Tyr701
KLH-conjugated synthetic peptide (C-DDPKRTG[pY]IKTE) corresponding to amino acids 694-705 of mouse STAT1, containing a N-terminal cysteine.

Application

Anti-phospho-STAT1 (Tyr701) Antibody is a Rabbit Polyclonal Antibody for detection of phospho-STAT1 (Tyr701) also known as Transcription factor ISGF-3 components p91/p84 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Biochem/physiol Actions

Other species cross-reactivity is unknown.
Phosphorylated STAT1

Physical form

Immunoaffinity Chromatography
Immunoaffinity purified IgG in 0.02 M PBS containing 0.1% Sodium Azide + 50% Glycerol and 5 mg/mL BSA.

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerol-containing solutions to become frozen during storage.

Analysis Note

Control
Mouse 3T3-L1 adipocytes stimulated with leukemia inhibitory factor (LIF).
Routinely evaluated by immunoblot in RIPA lysates from CNTF-treated SK-N-MC cells.

Western Blot Analysis:
A 1:500-1:2000 dilution of this lot detected phosphorylated STAT1 in RIPA lysates from CNTF-treated SK-N-MC cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
05-106404-88605-485
biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

mouse

antibody form

affinity purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

culture supernatant

antibody form

purified immunoglobulin

clone

polyclonal

clone

5C9.2, monoclonal

clone

A11W, monoclonal

clone

9E12, monoclonal

species reactivity

mouse, chicken, human, rat

species reactivity

rat, mouse, human

species reactivity

rat, chimpanzee, human, mouse, bovine

species reactivity

canine, human, mouse

shipped in

dry ice

shipped in

wet ice

shipped in

dry ice

shipped in

dry ice

Gene Information

human ... STAT1(6772)

Gene Information

human ... STAT1(6772)
mouse ... Stat1(20846)

Gene Information

bovine ... Stat5A(282375)

Gene Information

human ... STAT3(6774)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

The Janus kinase (JAK)/signal transducers and activators of transcription (STAT) signaling pathway plays an important role in cell proliferation, cell differentiation, cell migration, and cell death. It is the principal signaling mechanism for a variety of cytokines and growth factors. Constitutive activation or dysregulation of JAK/STAT signaling can result in inflammatory disease, erythrocytosis, gigantism, and leukemia.






Global Trade Item Number

SKUGTIN
07-30704053252325854

Questions

Reviews

No rating value

Active Filters