Skip to Content
Merck

Skip To

07-2113

Anti-phospho-Ribosomal Protein S6 Antibody (Ser240/Ser244)

from rabbit, purified by affinity chromatography

Synonym(s):

40S ribosomal protein S6, Phosphoprotein NP33

Sign In to View Organizational & Contract Pricing.

About This Item

eCl@ss:
32160702
UNSPSC Code:
12352203
NACRES:
NA.41
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
-


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), rat (based on 100% sequence homology), Xenopus (based on 100% sequence homology), mouse (based on 100% sequence homology), zebrafish (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer240/pSer244)

Gene Information

human ... RPS6(6194)

General description

S6 (Ribosomal protein S6, RpS6, 40S ribosomal protein S6) is known to play a role in cell growth and proliferation. It lays downstream of both the PI3K/Akt/mTOR pathway as a substrate of p70 S6 kinase and the Ras/Erk pathway as a substrate of RSK. S6 is phosphorylated in response to stimulation by growth factors, tumor promoting agents, and mitogens. It is dephosphorylated at growth arrest. There are two major areas of S6 phosphorylation; Ser235/Ser236 that are known to be phosphorylated by both p70S6 Kinase as well as RSK and Ser240/244 which is thought to only be phosphorylated by p70S6K.
~32 kDa observed

Immunogen

Epitope: Phosphorylated Ser240/Ser244
KLH-conjugated linear peptide corresponding to human Ribosomal Protein S6 phosphorylated at Ser240/Ser244.

Application

Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected Ribosomal Protein S6 in A431 cells and PMA treated A431 cells.
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling
This phospho-Ribosomal Protein S6 antibody is validated for use in WB & ICC for the detection of the phospho-Ribosomal Protein S6 protein.

Biochem/physiol Actions

This antibody recognizes Ribosomal Protein S6 phosphorylated at Ser240/Ser244.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
PMA treated and untreated A431 cell lysates
Evaluated by Western Blot in PMA treated and untreated A431 cell lysates.

Western Blot Analysis: 2 µg/mL of this antibody detected Ribosomal Protein S6 on 10 µg of PMA treated and untreated A431 cell lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
07-018-ISAB5500174MABS82
conjugate

unconjugated

conjugate

-

conjugate

unconjugated

conjugate

-

clone

polyclonal

clone

polyclonal

clone

SP45, monoclonal

clone

10G7.1, monoclonal

species reactivity

human

species reactivity

human

species reactivity

human (tested)

species reactivity

human, mouse

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

mouse

UniProt accession no.

P62753

UniProt accession no.

P23443

UniProt accession no.

P62753

UniProt accession no.

P23443

technique(s)

immunocytochemistry: suitable, western blot: suitable

technique(s)

inhibition assay: suitable (peptide), western blot: suitable

technique(s)

immunohistochemistry: 1:50

technique(s)

immunocytochemistry: suitable, western blot: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters