Skip to Content
Merck
All Photos(2)

Key Documents

06-1028

Sigma-Aldrich

Anti-Probable ergosterol biosynthetic protein 28 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Probable ergosterol biosynthetic protein 28, NET51

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€306.00

€306.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
€306.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€306.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rhesus macaque

species reactivity (predicted by homology)

rabbit (based on 100% sequence homology), orangutan (based on 100% sequence homology), mouse (based on 100% sequence homology), rhesus monkey (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ERG28(11161)

General description

Probable ergosterol biosynthetic protein 28, also known as NET51, is a very basic polypeptide containing several, evolutionary conserved transmembrane domains that are likely to play a role in sterol biosynthesis. It is found predominantly in normal testis and has also been found in many cancer cell lines and tumors.

Specificity

This antibody recognizes Probable ergosterol biosynthetic protein 28 near the first transmembrane domain.

Immunogen

Epitope: Transmembrane domain
KLH-conjugated linear peptide corresponding to human Probable ergosterol biosynthetic protein 28 near the first transmembrane domain.

Application

Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected Probable ergosterol biosynthetic protein 28 in HeLa cells.

Immunocytochemistry Analysis: A representative lot was used by an independent laboratory in rat liver cells. (Malik, P., et al. (2010). Cell. Mol. Life Sci. DOI 10.1007/s00018-010-0257-2.)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Hormones & Receptors
This Anti-Probable ergosterol biosynthetic protein 28 Antibody is validated for use in WB, IC for the detection of Probable ergosterol biosynthetic protein 28.

Quality

Evaluated by Western Blot in human testis tissue lysate.

Western Blot Analysis: 1:220 dilution of this antibody detected Probable ergosterol biosynthetic protein 28 on 10 µg of human testis tissue lysate.

Target description

~16 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human testis tissue lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service