Skip to Content
Merck

Skip To

07-1815

Anti-FIP200 Antibody

serum, from rabbit

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
1


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

rat ... Stx8(59074)

General description

200 kDa
Focal adhesion kinase family interacting protein of 200 kDa (FIP200) is a novel protein inhibitor for focal adhesion kinase (FAK). FIP200 inhibits kinase activity and associated cellular functions, such as cell adhesion, spreading, and motility in fibroblasts by binding directly to FAK.

Immunogen

Epitope: C-terminus
The immunogen was a synthetic peptide corresponding to the C-terminal region of FIP200.

Application

Detect FIP200 using this Anti-FIP200 Antibody validated for use in WB.
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling

Biochem/physiol Actions

This antibody recognizes the C-terminal region of FIP200.

Physical form

Unpurified rabbit serum in buffer containing 0.05% sodium azide.

Preparation Note

Stable for 2 years at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution.

Analysis Note

Evaluated by western blot on HEK293 cell lysate.
Western Blot Analysis: 1:1,000 dilution of this antibody was used to detect FIP200 in HEK293 cell lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABC128MABC114506-255-I
clone

polyclonal

clone

14E11.2, monoclonal

clone

73.3, monoclonal

clone

polyclonal

species reactivity

human, rat, mouse

species reactivity

human, mouse

species reactivity

mouse

species reactivity

human, rat, mouse

biological source

rabbit

biological source

mouse

biological source

mouse

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

serum

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

purified antibody

technique(s)

western blot: suitable

technique(s)

immunocytochemistry: suitable, western blot: suitable

technique(s)

flow cytometry: suitable

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters