Skip to Content
Merck

Skip To

04-775

Anti-HRAS Antibody

rabbit monoclonal, MC57

Synonym(s):

GTP- and GDP-binding peptide B, GTPase HRas, Ha-Ras1 proto-oncoprotein, Ras family small GTP binding protein H-Ras, Transforming protein p21, c-has/bas p21 protein, c-ras-Ki-2 activated oncogene, p19 H-RasIDX protein, transformation gene: oncogene HAMSV,

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 411,00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Clone:
MC57, monoclonal
Species reactivity:
rat, mouse, human
Application:
WB
Citations:
-

€ 411,00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1850

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-Ha-Ras Antibody, clone MC57, rabbit monoclonal, culture supernatant, clone MC57, from rabbit

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MC57, monoclonal

species reactivity

rat, mouse, human

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... HRAS(3265)

General description

Ras proteins are small GTP-binding proteins which, unlike the heterotrimeric G-proteins, contain all GTPase and effector functions within a single polypeptide. At least three isoforms of Ras exist, Ki-Ras, Ha-Ras and N-Ras, with distinct expression patterns but similar signaling activity. Ras is palmitoylated and farnesylated at the carboxy terminus, anchoring it in the membrane. In resting cells, Ras is loaded with GDP, and is activated subsequent to growth factor stimulation of receptors, which recruit Ras Guanine nucleotide Exchange Factors to the plane of the membrane. Proximity of exchange factors to the Ras proteins causes release of GDP, and its replacement by GTP. In its GTP-bound form, Ras binds several proteins, including Raf, RalGDS and PI3 Kinase. Inactivation of Ras occurs by GTP hydrolysis, which is greatly accelerated by RasGAP or NF-1, two known Ras GTPase Activating Proteins. It is possible to assay for Ras activation by incubation of lysates with the Ras-binding domain of Raf-1, which selectively binds to Ras:GTP.
~21 kDa

Immunogen

GST fusion protein corresponding to full length human Ha-Ras. Clone MC57.

Application

Detect Ha-Ras using this Anti-Ha-Ras Antibody, clone MC57 validated for use in WB.
Research Category
Signaling
Research Sub Category
MAP Kinases

Biochem/physiol Actions

Predicted to cross-react with mouse and rat based on sequence homology.
Recognizes Ha-Ras, Mr ~21 kDa. Does not cross react with Cdc42, Rho or Rac1.

Physical form

Cultured supernantant containing 0.05% sodium azide.

Preparation Note

Stable for 1 year at -20ºC from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for mingels.
Western Blot Analysis: A 1:1000 dilution of this antibody detected Ha-Ras in RIPA lysates from A431 cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 05-775

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
05-51616-321SAB4301113
antibody form

culture supernatant

antibody form

purified immunoglobulin

antibody form

purified antibody

antibody form

affinity isolated antibody

clone

MC57, monoclonal

clone

RAS10, monoclonal

clone

monoclonal

clone

polyclonal

biological source

rabbit

biological source

mouse

biological source

mouse

biological source

rabbit

Gene Information

human ... HRAS(3265)

Gene Information

human ... HRAS(3265), KRAS(3845), NRAS(4893)

Gene Information

human ... HRAS(3265), KRAS(3845), NRAS(4893)

Gene Information

human ... HRAS(3265)

species reactivity

rat, mouse, human

species reactivity

rat, human, mouse

species reactivity

mouse, human, rat

species reactivity

mouse, rat, human

technique(s)

western blot: suitable

technique(s)

ELISA: suitable, flow cytometry: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

western blot: 1:200-1:1000 (Cell Lysate)


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
04-77504053252677106

Questions

Reviews

No rating value

Active Filters