Skip to Content
Merck

Skip To

04-772

Anti-Src Antibody, CT, clone NL19 | 04-772

clone NL19, Upstate®, from rabbit

Synonym(s):

Proto-oncogene tyrosine-protein kinase SRC, Protooncogene SRC, Rous sarcoma tyrosine kinase pp60c-src, Tyrosine-protein kinase SRC-1, v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 566,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
NL19, monoclonal
Application:
IP, WB
Citations:
5

€ 566,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

NL19, monoclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SRC(6714)

General description

SRC-1 is a member of a gene family that functions in the full transcriptional activation of the steroid hormone receptor superfamily and contains intrinsic histone acetyltransferase activity. SRC-1 has a molecular weight of 160 kDa and has a basic helix-loop-helix (bHLH)-PAS domain.
~53-62kDa

Immunogen

KLH conjugated synthetic peptide (CKVADFGLARLIEDNEYTARQGA) of Src (avian sarcoma virus), residues 400-422. The immunogen sequence shares 22/23 amino acids with Yes and Fyn, and 21/23 amino acids with Lyn and Hck.

Application

Western Blot Analysis: A 1:2000 to 1:8000 dilution of this lot detected Src in RIPA whole cell lysates from human A431 cells. Immunoprecipitation: 5-20 μL of a previous lot immunoprecipitated Src from 500 μg of A431 RIPA lysate.

Biochem/physiol Actions

Predicted to cross react with frog and chicken. The immunogen sequence shares 22/23 amino acids with Yes and Fyn, and 21/23 amino acids with Lyn and Hck.
Src-tyrosine kinase family

Physical form

Format: Purified

Analysis Note

Control
RIPA whole cell lysates from human A431, Jurkat cells, mouse 3T3/A31 cells or rat PC-12 cells Included Positive Antigen Control: Catalog # 12-301, Non-Stimulated A431 cell lysate. Add 2.5 μL of 2-mercaptoethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.
routinely evaluated by immunoblot on RIPA whole cell lysates from human A431 and Jurkat cells, mouse 3T3/A31 cells or rat PC-12 cells

Other Notes

Replaces: 05-772

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
04-88905-67704-857
clone

NL19, monoclonal

clone

N6L, monoclonal

clone

9A6, monoclonal

clone

2N8, monoclonal

species reactivity

mouse, human, rat

species reactivity

human

species reactivity

mouse, human

species reactivity

vertebrates, mouse

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

conjugate

-

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

antibody form

culture supernatant

antibody form

culture supernatant

antibody form

purified immunoglobulin

antibody form

culture supernatant

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

western blot: suitable

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

western blot: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Machika Kawamura et al.
Life science alliance, 4(8) (2021-06-26)
The pericentromeric heterochromatin of one-cell embryos forms a unique, ring-like structure around the nucleolar precursor body, which is absent in somatic cells. Here, we found that the histone H3 variants H3.1 and/or H3.2 (H3.1/H3.2) were localized asymmetrically between the male
Pazit Y Cohen et al.
American journal of respiratory cell and molecular biology, 40(2), 231-238 (2008-08-05)
We have previously demonstrated that myofibroblasts from lungs with bleomycin-induced fibrosis overexpress FasL molecules. Two subpopulations of fibroblasts, distinguished by their expression of Thy1 molecules, have been shown in the lungs of both mice and humans. Thy1-mediated FasL induction has
Satoshi Funaya et al.
FEBS letters, 592(14), 2414-2424 (2018-07-03)
The chromatin structure in one-cell-stage mouse embryos is extremely loose and becomes tighter at the two-cell stage. As linker histones are involved in higher-order chromatin structure, we examined the involvement of the linker histone variant H1foo in the change of



Global Trade Item Number

SKUGTIN
04-77204053252402517

Questions

Reviews

No rating value

Active Filters