Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 200 μg | Check Cart for Availability | € 424,00 |
About This Item
€ 424,00
biological source
mouse
Quality Segment
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
7H4.2, monoclonal
species reactivity
human, rat, mouse
manufacturer/tradename
Upstate®
technique(s)
immunoprecipitation (IP): suitable, western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... MAPKAPK2(9261)
General description
Immunogen
Application
Immunoblot Analysis: 1:500
Immunoprecipitation: 1:20
Signaling
MAP Kinases
Kinases & Phosphatases
Physical form
Preparation Note
Analysis Note
Other Notes
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| species reactivity human, rat, mouse | species reactivity human, mouse | species reactivity rat, human, mouse | species reactivity human |
| clone 7H4.2, monoclonal | clone 3F6.5, monoclonal | clone UP-158-22-3-10, monoclonal | clone 14B9.1, monoclonal |
| biological source mouse | biological source mouse | biological source rabbit | biological source mouse |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate - |
| antibody form purified antibody | antibody form purified immunoglobulin | antibody form culture supernatant | antibody form purified immunoglobulin |
| shipped in wet ice | shipped in wet ice | shipped in dry ice | shipped in wet ice |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Storage Class
12 - Non Combustible Liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



