Skip to Content
Merck

Skip To

04-079

Anti-trimethyl-Histone H4 (Lys20), Antibody, rabbit monoclonal

culture supernatant, Upstate®

Synonym(s):

H4K20me3, Histone H4 (tri methyl K20)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 545,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
monoclonal
Species reactivity:
vertebrates, human
Application:
ChIP, DB, WB
Citations:
7

€ 545,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

monoclonal

species reactivity

vertebrates, human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable, dot blot: suitable, western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

trimethylation (Lys20)

Gene Information

human ... H4C1(8359)

General description

11kDa
Histones are nuclear proteins that form octameric structures which bind DNA to form units of chromatin called nucleosomes. The family of histones—H2A, H2B, H3, and H4—are key players in gene regulation. They undergo a number of post-translational modifications (PTM) in response to various stimuli, including phosphorylation on serine and threonine residues and methylation on lysine residues. PTMs produce configural changes in histone proteins that may induce nucleosome remodeling and expose or hide DNA sequences from transcriptional complexes. Histone H4 lysine 20 (H4K20) may undergo mono-, di-, or trimethylation, which is catalyzed by the methyltransferase PR-Set7 (Set8 or KMT5a). Methylated H4K20 plays a role in regulating DNA damage responses, mitosis, DNA replication, and gene expression. Trimethylation of H4K20 contributes to gene silencing, and is a mark of the repressive heterochromatin state.

Immunogen

Peptide corresponding to human Histone H4 containing the sequence [HRmethKVL] on which lysine 20 is trimethylated.

Application

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa cells (1 X 106 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 10 µL of either a negative control supernatant, or Anti-Trimethyl-Histone H4 (Lys20) antibody and the Magna ChIP A Kit (Cat. # 17-610). Successful immunoprecipitation of trimethyl-histone H4 (Lys20)-associated DNA fragments was verified by qPCR using ChIP Primers B-Globin. Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Dot Blot Analysis: Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665), which contain histone peptides with various modifications were probed with Cat. No 04-079, Anti-trimethyl Histone H4 (Lys20) (1:500 dilution). Proteins were visualized using a Donkey anti-rabbit IgG conjugated to HRP and a chemiluminescence detection system.
Detect trimethyl-Histone H4 (Lys20) using this Anti-trimethyl-Histone H4 (Lys20) Antibody, rabbit demontrated performance in WB, ChIP & DB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Biochem/physiol Actions

Recognizes Histone H4 when trimethylated on Lys20.

Physical form

100 μL of rabbit monoclonal IgG cell culture supernatant in 0.1% sodium azide

Preparation Note

2 years at -20°C from date of shipment

Analysis Note

Routinely evaluated by immunoblot.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
04-079-S07-46307-463-S
antibody form

culture supernatant

antibody form

culture supernatant

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Gene Information

human ... H4C1(8359)

Gene Information

human ... H4C1(8359)

Gene Information

human ... H4C1(8359)
mouse ... H4C1(326619)

Gene Information

human ... H4C1(8359)
mouse ... H4C1(326619)

species reactivity

vertebrates, human

species reactivity

human, vertebrates

species reactivity

human, mouse, rat

species reactivity

rat, human, mouse

clone

monoclonal

clone

monoclonal

clone

polyclonal

clone

polyclonal

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Cancer is a complex disease manifestation. At its core, it remains a disease of abnormal cellular proliferation and inappropriate gene expression. In the early days, carcinogenesis was viewed simply as resulting from a collection of genetic mutations that altered the gene expression of key oncogenic genes or tumor suppressor genes leading to uncontrolled growth and disease (Virani, S et al 2012). Today, however, research is showing that carcinogenesis results from the successive accumulation of heritable genetic and epigenetic changes. Moreover, the success in how we predict, treat and overcome cancer will likely involve not only understanding the consequences of direct genetic changes that can cause cancer, but also how the epigenetic and environmental changes cause cancer (Johnson C et al 2015; Waldmann T et al 2013). Epigenetics is the study of heritable gene expression as it relates to changes in DNA structure that are not tied to changes in DNA sequence but, instead, are tied to how the nucleic acid material is read or processed via the myriad of protein-protein, protein-nucleic acid, and nucleic acid-nucleic acid interactions that ultimately manifest themselves into a specific expression phenotype (Ngai SC et al 2012, Johnson C et al 2015). This review will discuss some of the principal aspects of epigenetic research and how they relate to our current understanding of carcinogenesis. Because epigenetics affects phenotype and changes in epigenetics are thought to be key to environmental adaptability and thus may in fact be reversed or manipulated, understanding the integration of experimental and epidemiologic science surrounding cancer and its many manifestations should lead to more effective cancer prognostics as well as treatments (Virani S et al 2012).


Volodymyr P Tryndyak et al.
Cancer biology & therapy, 5(1), 65-70 (2005-12-03)
Cancer cells are characterized by epigenetic dysregulation, including global genome hypomethylation, regional hypo- and hypermethylation, altered histone modifications, and disturbed genomic imprinting. Despite the long-established fact that global DNA hypomethylation is a common feature of tumors, very little is known
Dmitry Karachentsev et al.
Developmental biology, 304(1), 46-52 (2007-01-19)
Methylation of specific amino acids in histone tails is responsible for packaging DNA into condensed, repressed chromatin, and into open chromatin that is accessible to the transcription machinery. Monomethylation and trimethylation of histone H4-lysine 20 (H4-K20) control the formation of
Histone trimethylation and the maintenance of transcriptional ON and OFF states by trxG and PcG proteins.
Papp, B; Muller, J
Genes & Development null



Global Trade Item Number

SKUGTIN
04-07904053252624667

Questions

Reviews

No rating value

Active Filters