Skip to Content
Merck

Skip To

03-32-0009

Elastase Substrate VIII, Colorimetric

Substrate for the spectrophotometric assay of elastase.

Synonym(s):

Elastase Substrate VIII, Colorimetric

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
€ 226,00

About This Item

Empirical Formula (Hill Notation):
C19H25N5O8
CAS Number:
Molecular Weight:
451.43
UNSPSC Code:
12352204
NACRES:
NA.32
MDL number:

€ 226,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated

color

white to off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CCC(=O)O)C)C)C

InChI

1S/C19H25N5O8/c1-10(20-15(25)8-9-16(26)27)17(28)21-11(2)18(29)22-12(3)19(30)23-13-4-6-14(7-5-13)24(31)32/h4-7,10-12H,8-9H2,1-3H3,(H,20,25)(H,21,28)(H,22,29)(H,23,30)(H,26,27)/t10-,11-,12-/m0/s1

InChI key

GVUGADOWXGKRAE-SRVKXCTJSA-N

General description

Substrate for spectrophotometric assay of elastase.
Substrate for the spectrophotometric assay of elastase.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Substrate for the spectrophotometric assay of elastase
Product does not compete with ATP.
Reversible: no

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bieth, J., et al. 1974. Biochem. Med.11, 350.
Suc-Ala-Ala-Ala-pNA

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
324696324740324699
assay

≥98% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC), ≥95% (TLC)

assay

≥95% (HPLC)

solubility

DMSO: 5 mg/mL

solubility

water: 1 mg/mL

solubility

ethanol: 1 mg/mL, DMF: 5 mg/mL, DMSO: 5 mg/mL

solubility

ethanol: 1 mg/mL, water: soluble

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

form

solid

form

lyophilized

form

lyophilized solid, powder

form

lyophilized solid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters