Skip to Content
Merck
All Photos(1)

Documents

A-206

Supelco

Acylcarnitines Mix Stable Isotope Labeled solution

6-64 μg/mL (Methanol:1% aqueous formic acid (90:10) (v/v)), certified reference material, ampule of 0.5 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

Acetyl-L-carnitine (N-methyl-D3) inner salt, 64 μg/mL
Butyryl-L-Carnitine-(N-Methyl-D3), 6 μg/mL
L-Carnitine (trimethyl-D9) inner salt, 64 μg/mL
Decanoyl-L-Carnitine-(N-methyl-D3), 6 μg/mL
Glutaryl-L-Carnitine-(N-Methyl-D3) Lithium Salt, 15 μg/mL
Hexanoyl-L-Carnitine-(N-methyl-D3), 6 μg/mL
Isovaleryl-DL-Carnitine-(N,N,N-trimethyl-d9), 6 μg/mL
Lauroyl-L-Carnitine-(N,N,N-Trimethyl-D9), 6 μg/mL
Myristoyl-L-Carnitine-(N,N,N-trimethyl-D9), 6 μg/mL
Octanoyl-L-Carnitine-(N-Methyl-D3), 6 μg/mL
Palmitoyl-L-Carnitine (N-Methyl-D3), 20 μg/mL
Propionyl-L-Carnitine-(N-Methyl-D3), 10 μg/mL
Stearoyl-L-Carnitine-(N-methyl-D3), 12 μg/mL

packaging

ampule of 0.5 mL

manufacturer/tradename

Cerilliant®

concentration

6-64 μg/mL (Methanol:1% aqueous formic acid (90:10) (v/v))

format

multi-component solution

storage temp.

−70°C

General description

A single use multi-component stable isotope labeled internal standard mix of routinely tested Carnitine and Acetylcarnitines, suitable for the preparation of 250 mL DBS extraction solution sufficient for up to 2′500 Acylcarnitine test applications by MS/MS. The stable, quantitative solution format provides significant benefits to the user such as convenience of use, material savings, and increased efficiency of labor. Certified concentration values and lot-to-to consistency for each component provides increased efficiency from less time remaking pools due to dilution errors or rerunning samples. Acylcarnitines are biomarker for many inborn errors of metabolism that cause several metabolic diseases and are routinely tested in newborn screening programs.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - STOT SE 1

Target Organs

Eyes,Central nervous system

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service