Skip to Content
Merck
All Photos(1)

Key Documents

96378

Supelco

Stable Isotope Labeled Amino Acid Mix Solution 1

1250-2500 nmol/mL in 0.1 M HCl, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€606.00

€606.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€606.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.26

€606.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

liquid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

1250-2500 nmol/mL in 0.1 M HCl

technique(s)

HPLC: suitable
LC/MS: suitable

application(s)

clinical
food and beverages

format

multi-component solution

storage temp.

−20°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com


Check out the entire portfolio of our amino acids certified reference materials (CRM)

Analysis Note

17 amino acids in 0.1M hydrochloric acid
2.5 mM each: L-Alanine-13C3,15N, L-Arginine-13C6, L-Aspartic acid-13C4, L-Glutamic acid-13C5, Glycine-13C2,15N, L-Histidine-13C6, L-Isoleucine-13C6,15N, L-Leucine-13C6,15N, L-Lysine-13C6, L-Methionine-13C5,15N, L-Phenyl-13C6-alanine, L-Proline-13C5, L-Serine-13C3,15N, L-Threonine-13C4, L-Tyrosine-(phenyl-13C6), L-Valine-13C5
and 1.25mM L-Cystine-13C6,15N2

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service