Skip to Content
Merck
All Photos(1)

Key Documents

860844P

Avanti

C18 Galactosyl(β) Ceramide (d18:1/18:0)

Avanti Research - A Croda Brand 860844P, powder

Synonym(s):

D-galactosyl-β-1,1′-N-stearoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€220.00

€220.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€220.00

About This Item

Empirical Formula (Hill Notation):
C42H81 NO8
CAS Number:
Molecular Weight:
728.09
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€220.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860844P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860844P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@H]([C@H]1CO)O

InChI

1S/C42H81NO8/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-38(46)43-35(34-50-42-41(49)40(48)39(47)37(33-44)51-42)36(45)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h29,31,35-37,39-42,44-45,47-49H,3-28,30,32-34H2,1-2H3,(H,43,46)/b31-29+

InChI key

YMYQEDCYNANIPI-OWWNRXNESA-N

Application

C18 Galactosyl(β) Ceramide (d18:1/18:0) or D-galactosyl- β -1,1′-N-stearoyl-D-erythro-sphingosine has been used as a cerebroside standard in glycolipid isomer analysis using ultra-high resolution ion mobility spectrometry separations.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860844P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roza Wojcik et al.
International journal of molecular sciences, 18(1) (2017-01-21)
Understanding the biological roles and mechanisms of lipids and glycolipids is challenging due to the vast number of possible isomers that may exist. Mass spectrometry (MS) measurements are currently the dominant approach for studying and providing detailed information on lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service