Skip to Content
Merck
All Photos(1)

Key Documents

860598P

Avanti

C18 Lactosyl(β) Ceramide (d18:1/18:0)

Avanti Research - A Croda Brand 860598P, powder

Synonym(s):

D-lactosyl-β-1,1′ N-stearoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€220.00

€220.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€220.00

About This Item

Empirical Formula (Hill Notation):
C48H91NO13
CAS Number:
Molecular Weight:
890.24
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€220.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (860598P-25mg)
pkg of 1 × 5 mg (860598P-5mg)
pkg of 1 × 50 mg (860598P-50mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860598P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H]([C@H]1CO)O[C@H](O2)[C@H](O)[C@@H](O)[C@H]([C@H]2CO)O

InChI key

VOZHMDQUIRUFQW-LOTHNZFDSA-N

General description

C18 Lactosyl(β) Ceramide (d18:1/18:0) is a synthetic sphingolipid.[1]

Application

C18 Lactosyl (β) Ceramide (d18:1/18:0) is suitable for use as a lipid standard in ozone-induced dissociation mass spectrometry (OzID-MS).[2] It is also suitable for the preparation of saposin-derived lipid nanoparticles (SapNPs).[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860598P-25mg)
5 mL Amber Glass Screw Cap Vial (860598P-50mg)
5 mL Amber Glass Screw Cap Vial (860598P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceramide stearic to palmitic acid ratio predicts incident diabetes
Hilvo M, et al.
Diabetologia, 61(6), 1424-1434 (2018)
Saposin lipid nanoparticles: a highly versatile and modular tool for membrane protein research
Flayhan A, et al.
Structure, 26(2), 345-355 (2018)
Structural analysis of unsaturated glycosphingolipids using shotgun ozone-induced dissociation mass spectrometry
Barrientos RC, et al.
Journal of the American Society For Mass Spectrometry, 28(11), 2330-2343 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service