Skip to Content
Merck
All Photos(1)

Key Documents

860596P

Avanti

C18:1 Galactosyl(β) Ceramide (d18:1/18:1(9Z))

Avanti Research - A Croda Brand 860596P, powder

Synonym(s):

D-galactosyl-β-1,1′-N-oleoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€220.00

€220.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€220.00

About This Item

Empirical Formula (Hill Notation):
C42H79 NO8
CAS Number:
Molecular Weight:
726.08
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€220.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860596P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860596P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCC/C=C\CCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H](O)[C@H]1CO

InChI key

MVGFIPNJBNBHNC-VTRIZHIOSA-N

General description

Galactosyl ceramide is a complex glycosphingolipid.[1] It is a major component of the myelin sheath and contains a single galactose residue linked to ceramide.[1][2]Galactosyl ceramide is present mainly on the surface of colonic epithelial cells, vaginal epithelium and primary mammary epithelial cells.[3]

Biochem/physiol Actions

Galactosyl ceramide is essential for normal myelin function and stability.[4] It blocks epithelial cell-human immunodeficiency virus 1 (HIV-1) virion binding.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860596P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pediatr Neurol
Pediatric Neurology, 77(1), 306-318 (2013)
Gangliosides in Health and Disease
Gangliosides in Health and Disease, 77(1), 306-318 (2018)
K M Koshy et al.
Biophysical journal, 77(1), 306-318 (1999-07-02)
Divalent cations mediate a carbohydrate-carbohydrate association between the two major glycolipids, galactosylceramide (GalCer) and its sulfated form, cerebroside sulfate (CBS), of the myelin sheath. We have suggested that interaction between these glycolipids on apposed extracellular surfaces of myelin may be
S Moses Dennison et al.
Journal of virology, 88(16), 9406-9417 (2014-06-13)
Mucosal epithelial cell surface galactosylceramide (Galcer) has been postulated to be a receptor for HIV-1 envelope (Env) interactions with mucosal epithelial cells. Disruption of the HIV-1 Env interaction with such alternate receptors could be one strategy to prevent HIV-1 entry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service