Skip to Content
Merck
All Photos(2)

Key Documents

860431P

Avanti

C16 Galactosyl(α) Ceramide (d18:1/16:0)

D-galactosyl-α-1,1′ N-palmitoyl- D-erythro-sphingosine, powder

Synonym(s):

C16 α-D-galactosyl ceramide; N-(hexadecanoyl)-1-α-galactosyl-sphing-4-enine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€328.00

€328.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€328.00

About This Item

Empirical Formula (Hill Notation):
C40H77NO8
CAS Number:
Molecular Weight:
700.04
UNSPSC Code:
12352211
NACRES:
NA.25

€328.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (860431P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860431P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@]([C@@]([H])(/C=C/CCCCCCCCCCCCC)O)(NC(CCCCCCCCCCCCCCC)=O)CO[C@@H](O1)[C@H](O)[C@@H](O)[C@H]([C@H]1CO)O

InChI

1S/C40H77NO8/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(43)33(32-48-40-39(47)38(46)37(45)35(31-42)49-40)41-36(44)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h27,29,33-35,37-40,42-43,45-47H,3-26,28,30-32H2,1-2H3,(H,41,44)/b29-27+/t33-,34+,35+,37-,38-,39+,40-

InChI key

VJLLLMIZEJJZTE-KAQXTUTFSA-N

Application

C16 Galactosyl(α) Ceramide (d18:1/16:0) or α -GalCer (d18:1/16:0) is suitable for use as a hexosylceramides (HexCers) standard in high-performance thin layer chromatography (HPTLC).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860431P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathrin M Engel et al.
Biomolecules, 10(2) (2020-01-26)
The lipid composition of sperm membranes is crucial for fertilization and differs among species. As the evolution of internal fertilization modes in fishes is not understood, a comparative study of the sperm lipid composition in freshwater representatives of externally and
Diastereomer-specific quantification of bioactive hexosylceramides from bacteria and mammals
Von Gerichten J, et al.
Journal of Lipid Research, 58(6), 1247-1258 (2017)
Guang-Yi Dai et al.
The Journal of biological chemistry, 295(3), 717-728 (2019-12-11)
Cellular membranes contain many lipids, some of which, such as sphingolipids, have important structural and signaling functions. The common sphingolipid glucosylceramide (GlcCer) is present in plants, fungi, and animals. As a major plant sphingolipid, GlcCer is involved in the formation
Johanna von Gerichten et al.
Journal of lipid research, 58(6), 1247-1258 (2017-04-05)
Mammals synthesize, cell-type specifically, the diastereomeric hexosylceramides, β-galactosylceramide (GalCer) and β-glucosylceramide (GlcCer), which are involved in several diseases, such as sphingolipidosis, diabetes, chronic kidney diseases, or cancer. In contrast, Bacteroides fragilis, a member of the human gut microbiome, and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service