Skip to Content
Merck
All Photos(1)

Key Documents

860538P

Avanti

C8 Galactosyl(β) Ceramide (d18:1/8:0)

Avanti Research - A Croda Brand 860538P, powder

Synonym(s):

D-galactosyl-β-1,1′ N-octanoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€220.00

€220.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€220.00

About This Item

Empirical Formula (Hill Notation):
C32H61NO8
CAS Number:
Molecular Weight:
587.83
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€220.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (860538P-25mg)
pkg of 1 × 5 mg (860538P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860538P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@H]([C@H]1CO)O

InChI

1S/C32H61NO8/c1-3-5-7-9-10-11-12-13-14-15-16-18-19-21-26(35)25(33-28(36)22-20-17-8-6-4-2)24-40-32-31(39)30(38)29(37)27(23-34)41-32/h19,21,25-27,29-32,34-35,37-39H,3-18,20,22-24H2,1-2H3,(H,33,36)/b21-19+/t25-,26+,27?,29-,30?,31?,32+/m0/s1

InChI key

LCEXEEHGNKGJES-JLZVVTAUSA-N

General description

Galactosyl ceramide is a complex glycosphingolipid, which is a major component of the myelin sheath.[1] It consists of a single galactose residue linked to ceramide.[2]

Application

C8 Galactosyl(β) Ceramide (d18:1/8:0) may be suitable for the preparation of liposomes.[3][4]

Biochem/physiol Actions

Galactosyl ceramide is necessary for normal myelin function and stability.[5]

Packaging

5 mL Amber Glass Screw Cap Vial (860538P-25mg)
5 mL Amber Glass Screw Cap Vial (860538P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C8-glycosphingolipids preferentially insert into tumor cell membranes and promote chemotherapeutic drug uptake
Pedrosa L, et al.
Biochimica et Biophysica Acta - Biomembranes, 1848(8), 1656-1670 (2015)
Gangliosides in Health and Disease (2018)
Pediatric Neurology (2013)
Divalent cation-mediated interaction between cerebroside sulfate and cerebrosides: an investigation of the effect of structural variations of lipids by electrospray ionization mass spectrometry
Koshy KM, et al.
Biophysical Journal, 77(1), 306-318 (1999)
Biyu Hou et al.
Life sciences, 245, 117352-117352 (2020-02-02)
The depot-specific differences in lipidome of visceral adipose tissue (VAT) and subcutaneous adipose tissue (SAT) reflect heterogeneity of white adipose tissue (WAT), which plays a central role in its distinct response to outside stimuli. However, the detailed lipidome of depot-specific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service