Skip to Content
Merck
All Photos(1)

Key Documents

B3635

Sigma-Aldrich

Brain Extract from bovine brain

Type VII

Synonym(s):

Brain Extract from Bovine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€464.00

€464.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€464.00

About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

€464.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

bovine brain

type

Type VII

form

powder

lipid type

phosphoglycerides

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule.

Quality

Contains the major phospholipids and glycolipids normally found in brain tissue.

Preparation Note

A chloroform-methanol (1:2) extract washed with aqueous potassium chloride and precipitated with acetone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iven Winklemann et al.
The EMBO journal, 39(24), e105908-e105908 (2020-10-30)
Na+ /H+ exchangers (NHEs) are ancient membrane-bound nanomachines that work to regulate intracellular pH, sodium levels and cell volume. NHE activities contribute to the control of the cell cycle, cell proliferation, cell migration and vesicle trafficking. NHE dysfunction has been
Sharad Gupta et al.
The Journal of biological chemistry, 287(13), 9982-9989 (2011-12-22)
Huntington disease (HD) is one of several fatal neurodegenerative disorders associated with misfolded proteins. Here, we report a novel method for the sensitive detection of misfolded huntingtin (HTT) isolated from the brains of transgenic (Tg) mouse models of HD and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service