Skip to Content
Merck
All Photos(1)

Key Documents

860470P

Avanti

1-deoxysphingosine

Avanti Research - A Croda Brand 860470P, powder

Synonym(s):

1-deoxysphingosine (m18:1)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€237.00

€237.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€237.00

About This Item

Empirical Formula (Hill Notation):
C18H37NO
CAS Number:
Molecular Weight:
283.49
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€237.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860470P-1mg)
pkg of 1 × 10 mg (860470P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860470P

lipid type

bioactive lipids
sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@@](N)([H])[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI key

ZREXGWPJFRJAJU-SWESGNQDSA-N

General description

1-deoxysphingosine (1-deoxySO) has a (4E) double bond in its structure and is synthesized by the catabolism of 1-deoxyceramide in the presence of ceramidase enzyme.[1] It is an unsaturated deoxy-sphingoid base.[2]

Application

1-deoxysphingosine may be used for the complex preparation with bovine serum albumin for cytotoxicity testing of MN9D dopaminergic neuroblastoma cell line[3] and dorsal root ganglion neurons cultures.[2] It may also be used as a standard for quantification of spingolipids from human plasma samples by liquid chromatography electrospray ionization tandem mass spectrometry (LC−ESI−MS/MS).[4]

Biochem/physiol Actions

1-deoxysphingosine levels are elevated in lymphoblasts in hereditary sensory neuropathy type 1 (HSAN1) disorder.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (860470P-10mg)
5 mL Amber Glass Screw Cap Vial (860470P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regula Steiner et al.
Journal of lipid research, 57(7), 1194-1203 (2016-05-12)
The 1-deoxysphingolipids (1-deoxySLs) are formed by an alternate substrate usage of the enzyme, serine-palmitoyltransferase, and are devoid of the C1-OH-group present in canonical sphingolipids. Pathologically elevated 1-deoxySL levels are associated with the rare inherited neuropathy, HSAN1, and diabetes type 2
Analysis of sphingolipids in extracted human plasma using liquid chromatography electrospray ionization tandem mass spectrometry
Bui HH, et al.
Analytical Biochemistry, 423(2), 187-194 (2012)
Elucidating the chemical structure of native 1-deoxysphingosine
Steiner R, et al.
Journal of Lipid Research, 57(7), 1194-1203 (2016)
M F Dohrn et al.
European journal of neurology, 22(5), 806-814 (2015-01-28)
Diabetic distal sensorimotor polyneuropathy (DSPN) is a frequent, disabling complication of diabetes mellitus. There is increasing evidence that sphingolipids play a role in insulin resistance and type 2 diabetes (T2DM). Whether neurotoxic 1-deoxy-sphingolipids are elevated in DSPN patients' plasma and
Ceramide sphingolipid signaling mediates Tumor Necrosis Factor (TNF)-dependent toxicity via caspase signaling in dopaminergic neurons
Martinez TN, et al.
Mol. Neurodegener., 7(1), 45-45 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service