Skip to Content
Merck
All Photos(2)

Key Documents

860456P

Avanti

N-C16-deoxysphingosine

N-palmitoyl-1-deoxysphingosine (m18:1/16:0), powder

Synonym(s):

N-hexadecanoyl-1-deoxysphing-4-enine (m18:1/16:0); N-C16-1-deoxyCer; 110994

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€304.00

€304.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€304.00

About This Item

Empirical Formula (Hill Notation):
C34H67NO2
CAS Number:
Molecular Weight:
521.90
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€304.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (860456P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860456P

lipid type

sphingolipids
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Amber Glass Screw Cap Vial (860456P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah T Pruett et al.
Journal of lipid research, 49(8), 1621-1639 (2008-05-24)
"Sphingosin" was first described by J. L. W. Thudichum in 1884 and structurally characterized as 2S,3R,4E-2-aminooctadec-4-ene-1,3-diol in 1947 by Herb Carter, who also proposed the designation of "lipides derived from sphingosine as sphingolipides." This category of amino alcohols is now

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service