Skip to Content
Merck
All Photos(1)

Key Documents

541602C

Avanti

Soy Extract Polar

Avanti Research - A Croda Brand

Synonym(s):

Soybean Polar Lipid Extract

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€184.00
500 MG
€562.00
1 G
€903.00

€184.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€184.00
500 MG
€562.00
1 G
€903.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€184.00


Check Cart for Availability

Request a Bulk Order

form

chloroform solution

packaging

pkg of 2 × 20 mL (541602C-1g)
pkg of 1 × 4 mL (541602C-100mg)
pkg of 5 × 4 mL (541602C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

25 mg/mL (541602C-100mg)
25 mg/mL (541602C-1g)
25 mg/mL (541602C-500mg)

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

Polar Lipid Extract is derived from the total lipid extract by precipitation with acetone followed by extraction of the acetone insoluble material with diethyl ether.
The Soy Extract Polar (SPE) comprises majorly phosphatidylcholine (45.7%), phosphatidylethanolamine (PE 22.1%) and phosphatidylinositol (PI, 18.4%). It also has phosphatidic acid (PA, 6.9%) and a mixture of other unknown lipids (6.9%).[1] Soybean is the natural source of soy polar extract.[2]

Application

Soy Extract Polar has been used:
  • in the preparation of liposomes for forester resonance energy transfer (FRET)-based assay[3]
  • along with 1,2-dioleoyl-sn-glycero-3-phospho-L-serine (sodium salt) (DOPS) to mimic the cell membrane inner leaflet and for the preparation of giant unilamellar vesicle (GUV) connected to a multilamellar vesicle (MLV)[1]
  • in liposome preparation for the generation of GUV-MLV reservoirs[4]

Biochem/physiol Actions

Soy Extract Polar (SPE) is useful in the preparation of unilamellar liposomes and also nanotube networks.[5] The hockey-puck-like structure of the SPE provides a native lipid membrane for various studies.[2]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (541602C-1g)
5 mL Clear Glass Sealed Ampule (541602C-100mg)
5 mL Clear Glass Sealed Ampule (541602C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Membrane remodeling of giant vesicles in response to localized calcium ion gradients
Doosti BA
Journal of Visualized Experiments, e57789-e57789 (2018)
A cell-penetrating scorpion toxin enables mode-specific modulation of TRPA1 and pain
King JV, et al.
Cell, 178(6), 1362-1374 (2019)
Short-chain alcohols promote accelerated membrane distention in a dynamic liposome model of exocytosis
Wittenberg NJ, et al.
Langmuir, 24(6), 2637-2642 (2008)
Johan Berg et al.
Biochimica et biophysica acta. Bioenergetics, 1861(2), 148116-148116 (2019-11-17)
Data from earlier studies showed that minor structural changes at the surface of cytochrome c oxidase, in one of the proton-input pathways (the D pathway), result in dramatically decreased activity and a lower proton-pumping stoichiometry. To further investigate how changes
Generation of phospholipid vesicle-nanotube networks and transport of molecules therein
Jesorka A, et al.
Nature Protocols, 6(6), 791-791 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service