Skip to Content
Merck
All Photos(1)

Key Documents

860533P

Avanti

C16 Ceramide-1-Phosphate (d18:1/16:0)

Avanti Research - A Croda Brand 860533P, powder

Synonym(s):

N-palmitoyl-ceramide-1-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€181.00

€181.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€181.00

About This Item

Empirical Formula (Hill Notation):
C34H71N2O6P
CAS Number:
Molecular Weight:
634.91
UNSPSC Code:
12352211
NACRES:
NA.25

€181.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860533P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860533P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCC)=O)COP([O-])(O)=O.[NH4+]

InChI

1S/C34H68NO6P.H3N/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-33(36)32(31-41-42(38,39)40)35-34(37)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h27,29,32-33,36H,3-26,28,30-31H2,1-2H3,(H,35,37)(H2,38,39,40);1H3/b29-27+;/t32-,33+;/m0./s1

InChI key

RCRVTEZMCBPWGN-YWBCJBABSA-N

General description

C16 Ceramide-1-Phosphate (d18:1/16:0) or N-palmitoyl-ceramide-1-phosphate is a natural long chain ceramide.[1]

Application

C16 Ceramide-1-Phosphate (d18:1/16:0) or N-palmitoyl-ceramide-1-phosphate has been used in lipid preparation to treat endothelial colony-forming cells (ECFCs) for nuclear morphology analysis.[1] It has also been used as a bioactive lipid in cell migration assay to study its effects on multipotent stromal cell (MSCs) and human umbilical vein endothelial cells (HUVECs).[2]

Packaging

5 mL Amber Glass Screw Cap Vial (860533P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chihwa Kim et al.
Stem cells (Dayton, Ohio), 31(3), 500-510 (2012-11-30)
Ceramide-1-phosphate (C1P) is a bioactive lipid that, in contrast to ceramide, is an antiapoptotic molecule released from cells that are damaged and "leaky." As reported recently, C1P promotes migration of hematopoietic cells. In this article, we tested the hypothesis that
Hebe Agustina Mena et al.
Arteriosclerosis, thrombosis, and vascular biology, 39(10), e219-e232 (2019-08-23)
Ceramide 1-phosphate (C1P) is a bioactive sphingolipid highly augmented in damaged tissues. Because of its abilities to stimulate migration of murine bone marrow-derived progenitor cells, it has been suggested that C1P might be involved in tissue regeneration. In the present
Antonio Gómez-Muñoz et al.
Journal of lipid research, 45(1), 99-105 (2003-10-03)
It was reported previously that ceramide-1-phosphate (Cer-1-P) is mitogenic for fibroblasts (Gómez-Muñoz, A., P. A. Duffy, A. Martin, L. O'Brien, H-S. Byun, R. Bittman, and D. N. Brindley. 1995. Mol. Pharmacol. 47: 883-889; Gómez-Muñoz, A., L. M. Frago, L. Alvarez
V T Hinkovska-Galcheva et al.
The Journal of biological chemistry, 273(50), 33203-33209 (1998-12-05)
Ceramide, a product of agonist-stimulated sphingomyelinase activation, is known to be generated during the phagocytosis of antibody-coated erythrocytes by polymorphonuclear leukocytes. Agonist-stimulated formation of ceramide-1-phosphate is now shown to occur in 32PO4-labeled neutrophils. Ceramide-1-phosphate is formed by a calcium-dependent ceramide
A Gomez-Muñoz et al.
Molecular pharmacology, 47(5), 833-839 (1995-05-01)
Ceramide and ceramide-1-phosphate are sphingolipid analogues of diacylglycerol and phosphatidate, respectively, and they are putative second messengers of agonist-stimulated sphingomyelin metabolism. The interactions of exogenous cell-permeable ceramides and ceramide-1-phosphates in modifying DNA synthesis and signal transduction were investigated in Rat-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service