Skip to Content
Merck
All Photos(2)

Key Documents

860361P

Avanti

18:0 PC-d4

Avanti Research - A Croda Brand 860361P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphocholine-1,1,2,2-d4

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€1,000.00

€1,000.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€1,000.00

About This Item

Empirical Formula (Hill Notation):
C44H84NO8PD4
CAS Number:
Molecular Weight:
794.17
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

€1,000.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860361P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860361P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC([2H])([2H])C([2H])([2H])[N+](C)(C)C)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O

InChI key

NRJAVPSFFCBXDT-HFCWBTDDSA-N

General description

18:0 PC-d4 is a deuterated phosphocholine (PC) in which four protons of distearoyl are replaced by deuterium.

Application

18:0 PC-d4 has been used as a deuterated phospholipid (PL) standard for the quantification of lipid from urine samples using liquid chromatography-mass spectrometry (LC-MS).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860361P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biomarkers to monitor drug-induced phospholipidosis
Baronas ET, et al.
Toxicology and Applied Pharmacology, 218(1), 72-78 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service