Skip to Content
Merck
All Photos(1)

Key Documents

860900C

Avanti

14:0-16:1-14:0 D5 TG

Avanti Research - A Croda Brand 860900C

Synonym(s):

1,3(d5)-ditetradecanoyl-2-(9Z-hexadecenoyl)-glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H83D5O6
CAS Number:
Molecular Weight:
754.23
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.12

form

liquid

packaging

pkg of 1 × 1 mL (860900C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860900C

concentration

1 mg/mL (860900C-1mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[2H]C(C([2H])([2H])OC(CCCCCCCCCCCCC)=O)(OC(CCCCCCC/C=C\CCCCCC)=O)C([2H])([2H])OC(CCCCCCCCCCCCC)=O

InChI key

JJUUQTPTIFXQFZ-RZCIEKIASA-N

General description

14:0-16:1-14:0 D5 TG, also known as 1,3(d5)-ditetradecanoyl-2-(9Z-hexadecenoyl)-glycerol, belongs to the class of prostaglandin deuterated isotopes.

Application

14:0-16:1-14:0 D5 TG or 1,3(d5)-ditetradecanoyl-2-(9Z-hexadecenoyl)-glycerol has been used as an internal standard in mass spectrometry analysis of triacylglycerides. It has also been used as an internal standard in the extraction protocol to separate neutral lipids from polar lipids.

Packaging

5 mL Clear Glass Sealed Ampule (860900C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weiqin Chen et al.
PloS one, 8(12), e82526-e82526 (2013-12-21)
Mutations in BSCL2 underlie human congenital generalized lipodystrophy type 2 disease. We previously reported that Bscl2 (-/-) mice develop lipodystrophy of white adipose tissue (WAT) due to unbridled lipolysis. The residual epididymal WAT (EWAT) displays a browning phenotype with much
Methods in Enzymology. Methods of adipose tissue biology, part B. Preface.
Ormond A MacDougald
Methods in enzymology, 538, xv-xv (2014-02-18)
Jeff D Moore et al.
Methods in enzymology, 432, 351-367 (2007-10-24)
Qualification, preparation, and use of lipid compounds as analytical reference standards are daunting endeavors. The sheer vastness of the number of lipid compounds present in biological samples make it impossible to directly standardize each entity. Available lipid compounds chosen for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service