Skip to Content
Merck
All Photos(3)

Key Documents

860061P

Avanti

Egg SM

Avanti Research - A Croda Brand

Synonym(s):

16:0 SM; Hexadecanoyl Sphingomyelin; N-hexadecanoyl-D-erythro-sphingosylphosphorylcholine; N-(hexadecanoyl)-sphing-4-enine-1-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€151.00
200 MG
€297.00
1 G
€719.00

€151.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€151.00
200 MG
€297.00
1 G
€719.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€151.00


Check Cart for Availability

Request a Bulk Order

description

Sphingomyelin (Egg, Chicken)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (860061P-1g)
pkg of 1 × 200 mg (860061P-200mg)
pkg of 1 × 25 mg (860061P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCC)=O)COP([O-])(OCC[N+](C)(C)C)=O

InChI

1S/C39H79N2O6P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-38(42)37(36-47-48(44,45)46-35-34-41(3,4)5)40-39(43)33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h30,32,37-38,42H,6-29,31,33-36H2,1-5H3,(H-,40,43,44,45)/b32-30+/t37-,38+/m0/s1

InChI key

RWKUXQNLWDTSLO-GWQJGLRPSA-N

General description

As a major constituent of cell membranes, sphingomyelin is found at particularly high concentrations in the membranes of nerve cells (in the myelin sheaths) and red blood cells. It was previously thought to have a purely structural role, similar to the function of phosphatidylcholine, through intermolecular interactions mediated by the 2-amide group, the 3-hydroxy group and the 4,5-trans double bond of the sphingoid base.[1] However, it is now appreciated that sphingomyelin has a high affinity for cholesterol and that these two lipids pack tightly into liquid-ordered domains among a liquid-disordered phase to form lipid rafts.[1] These membrane microdomains are thought to function as signaling platforms that regulate the localization and interactions of proteins. But sphingomyelin does not just influence signaling as a component of lipid rafts — it is also a precursor to ceramides and other sphingolipid metabolites that comprise the sphingomyelin cycle or sphingolipid network.[1]
Sphingomyelin and related lipids

Application

Egg SM has been used for the preparation of sphingomyelin (SM) standard solutions.[2] It has also been used in the preparation of liposomes.[3][4]

Packaging

20 mL Clear Glass Screw Cap Vial (860061P-1g)
5 mL Amber Glass Screw Cap Vial (860061P-200mg)
5 mL Amber Glass Screw Cap Vial (860061P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Visualization of the two-step fusion process of the retrovirus avian sarcoma/leukosis virus by cryo-electron tomography
Cardone G, et al.
Journal of Virology, 86(22), 12129-12137 (2012)
Maia Kinnebrew et al.
eLife, 8 (2019-10-29)
Previously we proposed that transmission of the hedgehog signal across the plasma membrane by Smoothened is triggered by its interaction with cholesterol (Luchetti et al., 2016). But how is cholesterol, an abundant lipid, regulated tightly enough to control a signaling
Protocols for Enzymatic Fluorometric Assays to Quantify Phospholipid Classes
Morita S, et al.
International Journal of Molecular Sciences, 21(3), 1032-1032 (2020)
Yuanqing Ma et al.
Biophysical journal, 115(8), 1498-1508 (2018-10-03)
Membrane viscosity and hydration levels characterize the biophysical properties of biological membranes and are reflected in the rate and extent of solvent relaxation, respectively, of environmentally sensitive fluorophores such as Laurdan. Here, we first developed a method for a time-resolved
Takaaki Matsufuji et al.
Langmuir : the ACS journal of surfaces and colloids, 34(1), 465-471 (2017-12-13)
Ceramide is a bioactive lipid with important roles in several biological processes including cell proliferation and apoptosis. Although 3-ketoceramides that contain a keto group in place of the 3-OH group of ceramide occur naturally, ceramide derivatives oxidized at the primary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service