Skip to Content
Merck
All Photos(3)

Key Documents

840021P

Avanti

Egg PE

Avanti Research - A Croda Brand 840021P

Synonym(s):

L-α-phosphatidylethanolamine (Egg, Chicken)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€480.00
200 MG
€1,410.00
500 MG
€2,080.00

€480.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€480.00
200 MG
€1,410.00
500 MG
€2,080.00

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

€480.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (840021P-1g)
pkg of 2 × 100 mg (840021P-200mg)
pkg of 5 × 100 mg (840021P-500mg)
pkg of 1 × 25 mg (840021P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840021P

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O

InChI

1S/C23H48NO7P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-23(26)29-20-22(25)21-31-32(27,28)30-19-18-24/h22,25H,2-21,24H2,1H3,(H,27,28)/t22-/m1/s1

InChI key

BBYWOYAFBUOUFP-JOCHJYFZSA-N

General description

Egg PE is a natural phosphatidylethanolamine (PE).[1]

Application

Egg PE has been used for the preparation of glycosphingolipids (GSL)-containing liposomes.[1] It has also been used to prepare phosphatidylethanolamine stock solution.[2]

Packaging

5 mL Clear Glass Sealed Ampule (840021P-200mg)
5 mL Clear Glass Sealed Ampule (840021P-25mg)
5 mL Clear Glass Sealed Ampule (840021P-500mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (840021P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liposome Methods and Protocols (2002)
Plant Lipid Signaling Protocols (2013)
Bixia Zhang et al.
Plant physiology (2020-04-26)
Cinnamate 4-hydroxylase (C4H, CYP73A) is a cytochrome P450 monooxygenase associated externally with endoplasmic reticulum of plant cells. The enzyme uses NADPH-cytochrome P450 reductase (CPR) as a donor of electrons and hydroxylates cinnamic acid to form 4-coumaric acid in phenylpropanoid metabolism.
Małgorzata A Przeradzka et al.
Journal of thrombosis and haemostasis : JTH, 18(2), 364-372 (2019-11-02)
The identity of the amino acid regions of factor VIII (FVIII) that contribute to factor IXa (FIXa) and von Willebrand factor (VWF) binding has not been fully resolved. Previously, we observed that replacing the FVIII C1 domain for the one
Daichi Yamaura et al.
Langmuir : the ACS journal of surfaces and colloids, 34(19), 5615-5622 (2018-04-18)
Artificial bilayer lipid membranes (BLMs) provide well-defined systems for investigating the fundamental properties of membrane proteins, including ion channels, and for screening the effect of drugs that act on them. However, the application of this technique is limited due to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service