Skip to Content
Merck
All Photos(3)

Key Documents

841118P

Avanti

Egg Trans PE

L-α-phosphatidylethanolamine, transphosphatidylated (Egg, Chicken), powder

Synonym(s):

L-a-Phosphatidylethanolamine Transphosphatidylated (prepared from Egg PC)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€133.00

€133.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€133.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€133.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (841118P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 841118P

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C39H76NO8P/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-39(42)48-37(36-47-49(43,44)46-34-33-40)35-45-38(41)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h17-18,37H,3-16,19-36,40H2,1-2H3,(H,43,44)/b18-17-/t37-/m1/s1

InChI key

FHQVHHIBKUMWTI-OTMQOFQLSA-N

General description

Phosphatidylethanolamine (PE) is a phospholipid found in all living organisms.[1] The phosphatidylethanolamine (PE) headgroup is a zwitterion, with a negatively charged phosphate group and positively charged free amine group.[2] The egg yolk contains high levels of unsaturated PE which accounts to 83.10%.[3]

Application

Egg Trans PE is suitable for use:
  • in the preparation of zwitterionic liposome for binding assay[4]
  • as a component in optimized theranostic nanoemulsion (NE) formulations[5]
  • in the preparation of large unilamellar vesicles (LUVs) for detecting the leakage of intraliposomal carboxyfluorescein caused by anthraquinones[6]

Biochem/physiol Actions

Phosphatidylethanolamine (PE) along with other phospholipids forms the backbone of biological membranes. PE is known to regulate autophagy. The autophagic membranes possesses PE with high degree of unsaturation.[1] Egg yolk PE has high radical scavenging activity owing to antioxidant potential.[3]

Packaging

5 mL Clear Glass Sealed Ampule (841118P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niravkumar R Patel et al.
Cancer biology & therapy, 19(7), 554-564 (2018-05-09)
Ovarian cancer ranks fifth in cancer related deaths for women in USA. The high mortality rate associated with ovarian cancer is due to diagnosis at later stages of disease and the high recurrence rate of 60-80%. Recurrent ovarian cancers are
Membrane permeabilization by trypanosome lytic factor, a cytolytic human high density lipoprotein
Harrington JM, et al.
Journal of biological and chemical chronicles, 284(20), 13505-13512 (2009)
The
The Membranes of Cells, 22(3) (2016)
Egg Yolk Phosphatidylethanolamine: Extraction Optimization, Antioxidative Activity, and Molecular Structure Profiling
Sun N, et al.
Journal of Food Science, 84(5), 1002-1011 (2019)
In Vitro and In Vivo evaluation of a novel folate-targeted theranostic nanoemulsion of docetaxel for imaging and improved anticancer activity against ovarian cancers
Patel NR, et al.
Cancer Biochemistry Biophysics, 19(7), 554-564 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service