Skip to Content
Merck
All Photos(2)

Key Documents

852472C

Avanti

C18(Plasm)-22:6 PC

1-(1Z-octadecenyl)-2-docosahexaenoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-O-1′-(Z)-octadecenyl-2-(4Z,7Z,10Z,13Z,16Z,19Z-docosahexaenoyl)-sn-glycero-3-phosphocholine; GPCho(18:0p/22:6(4Z,7Z,10Z,13Z,16Z,19Z))

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€276.00

€276.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€276.00

About This Item

Empirical Formula (Hill Notation):
C48H84NO7P
CAS Number:
Molecular Weight:
818.16
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€276.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 5 mL (852472C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 852472C

concentration

1 mg/mL (852472C-5mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

Application

C18(Plasm)-22:6 PC has been used in miscibility studies.[1] It has also been used for quantification of phosphotidylcholine in total lipid extracts of bovine retina.[2]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (852472C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Identification and quantification of phosphatidylcholines containing very-long-chain polyunsaturated fatty acid in bovine and human retina using liquid chromatography/tandem mass spectrometry
Berdeaux O, et al.
Journal of Chromatography A, 1217(49), 7738-7748 (2010)
Organization of lipids in the artificial outer membrane of bull spermatozoa reconstructed at the air-water interface
Le Guillou J, et al.
Colloids and Surfaces. B, Biointerfaces, 108, 246-254 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service