Skip to Content
Merck
All Photos(2)

Key Documents

852806C

Avanti

C18(Plasm)-22:6 PE

1-(1Z-octadecenyl)-2-docosahexaenoyl-sn-glycero-3-phosphoethanolamine, chloroform

Synonym(s):

1-O-1′-(Z)-octadecenyl-2-(4Z,7Z,10Z,13Z,16Z,19Z-docosahexaenoyl)-sn-glycero-3-phosphoethanolamine; GPEtn(18:0p/22:6(4Z,7Z,10Z,13Z,16Z,19Z))

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€543.00
25 MG
€2,590.00

€543.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€543.00
25 MG
€2,590.00

About This Item

Empirical Formula (Hill Notation):
C45H78NO7P
CAS Number:
Molecular Weight:
776.08
UNSPSC Code:
51191904
NACRES:
NA.25

€543.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 5 mL (852806C-25mg)
pkg of 1 × 5 mL (852806C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 852806C

concentration

1 mg/mL (852806C-5mg)
5 mg/mL (852806C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O)CO/C=C\CCCCCCCCCCCCCCCC

InChI key

FIJFPUAJUDAZEY-MNDXXDKYSA-N

Application

C18(Plasm)-22:6 PE has been used as a standard in liquid chromatography−mass spectrometry (LC-MS) method[1]. It has also been used as a component of plasmalogen in cholesterol efflux capacity studies.[2]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (852806C-25mg)
30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (852806C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipid composition of the human eye: are red blood cells a good mirror of retinal and optic nerve fatty acids?
Acar N, et al.
Testing, 7(4) (2012)
Association of cholesterol efflux capacity with plasmalogen levels of high-density lipoprotein: A cross-sectional study in chronic kidney disease patients
Maeba R, et al.
Atherosclerosis, 270(4), 102-109 (2018)
Lipid composition of the human eye: are red blood cells a good mirror of retinal and optic nerve fatty acids?
Acar N, et al.
PLoS ONE, 7(4) (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service