Skip to Content
Merck
All Photos(1)

Key Documents

840527P

Avanti

Hydrogenated DGDG

Avanti Research - A Croda Brand, powder

Synonym(s):

Digalactosyldiacylglycerol (plant, hydrogenated)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€393.00

€393.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€393.00

About This Item

Empirical Formula (Hill Notation):
C51H96O15
CAS Number:
Molecular Weight:
949.30
UNSPSC Code:
12352211
NACRES:
NA.25

€393.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (840527P-10MG)
pkg of 1 × 5 mg (840527P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](COC(CCCCCCCCCCCCCCCCC)=O)(OC(CCCCCCCCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@H]([C@H]1COC(O2)[C@H](O)[C@@H](O)[C@H]([C@H]2CO)O)O

InChI

1S/C51H96O15/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-42(53)61-36-39(64-43(54)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)37-62-50-49(60)47(58)45(56)41(66-50)38-63-51-48(59)46(57)44(55)40(35-52)65-51/h39-41,44-52,55-60H,3-38H2,1-2H3/t39-,40-,41-,

InChI key

LDQFLSUQYHBXSX-SIGZKIDOSA-N

General description

Digalactosyldiacylglycerol (DGDG) is a galactosylglycerolipid, which is a major lipid component in the thylakoid membrane of leaf chloroplasts. It is also present in the fruits, grains and roots.[1] DGDG is synthesized from Monogalactosyldiacylglycerol (MGDG) by the enzyme DGDG synthase DGD1 .[2]

Biochem/physiol Actions

Digalactosyldiacylglycerol (DGDG) along with monogalactosyldiacylglycerol (MGDG) is involved in chloroplast membrane biogenesis and photosynthesis. DGDG and MGDG are responsible for the intake of proteins into the chloroplast.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (840527P-5MG and 840527P-10MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The digalactosyldiacylglycerol (DGDG) synthase DGD1 is inserted into the outer envelope membrane of chloroplasts in a manner independent of the general import pathway and does not depend on direct interaction with monogalactosyldiacylglycerol synthase for DGDG biosynthesis
Froehlich J E, et al.
The Journal of biological chemistry, 276(34), 31806-31812 (2001)
The galactolipid monogalactosyldiacylglycerol (MGDG) contributes to photosynthesis-related processes in Arabidopsis thaliana
Aronsson H
Plant signaling & behavior, 3(12), 1093-1095 (2008)
Digestion of plant monogalactosyldiacylglycerol and digalactosyldiacylglycerol in rat alimentary canal
Sugawara T and Miyazawa T
The Journal of Nutritional Biochemistry, 11(3), 147-152 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service