Skip to Content
Merck
All Photos(2)

Key Documents

857464P

Avanti

DGTS

Avanti Research - A Croda Brand

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-O-4′-(N,N,N-trimethyl)-homoserine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€161.00
5 MG
€364.00

€161.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€161.00
5 MG
€364.00

About This Item

Empirical Formula (Hill Notation):
C42H81NO7
CAS Number:
Molecular Weight:
712.10
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€161.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (857464P-1mg)
pkg of 1 × 5 mg (857464P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phosphoglycerides
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COCCC([N+](C)(C)C)C([O-])=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C42H81NO7/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-40(44)49-37-38(36-48-35-34-39(42(46)47)43(3,4)5)50-41(45)33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h38-39H,6-37H2,1-5H3/t38-,39?/m0/s1

InChI key

JENDGLMWMJVGPR-SQTCDLSVSA-N

General description

Diacylglyceryltrimethylhomo-Ser (DGTS) is an abundant lipid in the membranes of many algae, lower plants, and fungi. It commonly has an inverse concentration relationship with phosphatidylcholine, thus seemingly capable of replacing this phospholipid in these organisms.

Application

Diacylglyceryl-N,N,N-trimethylhomoserine (DGTS) is suitable for use as a lipid standard in thin layer chromatography (TLC).[1][2]

Packaging

5 mL Clear Glass Sealed Ampule (857464P-1mg)
5 mL Clear Glass Sealed Ampule (857464P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cellular tracking and gene profiling of Fusarium graminearum during maize stalk rot disease development elucidates its strategies in confronting phosphorus limitation in the host apoplast
Zhang Y, et al.
PLoS Pathogens, 12(3), e1005485-e1005485 (2016)
The PHO signaling pathway directs lipid remodeling in Cryptococcus neoformans via DGTS synthase to recycle phosphate during phosphate deficiency
Lev S, et al.
PLoS ONE, 14(2), e0212651-e0212651 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service